Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565473_m1
          See other IGFBP2 GE Assays ›
          Gene Symbol:
          Igfbp2
          Gene Name:
          insulin-like growth factor binding protein 2
          Gene Aliases:
          IGFBP-2, ILGFBPA
          Chromosome Location:
          Chr.9: 80118029 - 80144804 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565473_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igfbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013122.2 NP_037254.2 2 - 3 678 93
          GenBank mRNA BC092570.1 - 2 - 3 678 7308779 93
          FQ212531.1 - 2 - 3 697 93
          J04486.1 - 2 - 3 870 93
          M31672.1 - 2 - 3 706 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000874.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igfbp2

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          signal transduction
          multicellular organism development
          female pregnancy
          aging
          response to nutrient
          response to mechanical stimulus
          response to lithium ion
          response to estradiol
          response to retinoic acid
          cellular response to hormone stimulus
          positive regulation of activated T cell proliferation
          response to drug
          regulation of insulin-like growth factor receptor signaling pathway
          response to estrogen
          response to steroid hormone
          response to glucocorticoid
          insulin-like growth factor binding
          insulin-like growth factor I binding
          insulin-like growth factor II binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P039026 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56703 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103424 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115922 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039026 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032415 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039026 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032415 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032415 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P039026 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P547185 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700145663 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50638 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33179 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66535 - -
          Agilent Rat cDNA Microarray (G4105A) 700607330 - -
          Agilent Rat cDNA Microarray (G4105A) 700145663 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10924227 - -
          Affymetrix Rat Gene 1.0 ST Array 10924225 - -
          Affymetrix Rat Gene 1.0 ST Array 10924228 - -
          Affymetrix Rat Gene 1.0 ST Array 10924224 - -
          Affymetrix Rat Gene 1.0 ST Array 10924226 - -
          Affymetrix Rat Gene 1.0 ST Array 10924229 - -
          Affymetrix Rat Gene 1.1 ST Array 10924223 - -
          Rat Genome 230 2.0 Array 1386878_PM_at - -
          Rat Genome 230 2.0 Array 1367648_PM_at - -
          Rat Genome 230 2.0 Array 1367648_at - -
          Rat Genome 230 2.0 Array 1386878_at - -
          Affymetrix HT Rat Focus Array 1367648_at - -
          Affymetrix HT Rat Focus Array 1367648_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900007170.rn.1 TC0900000874.rn.1 RTA TC
          Rat Expression Set 230 1367648_PM_at - -
          Rat Expression Set 230 1367648_at - -
          Affymetrix Rat Gene 1.0 ST Array 10924223 - -
          Rat Genome U34 Set A09811cds_s_at - -
          Rat Genome U34 Set M91595exon_s_at - -
          Rat Genome U34 Set J04486_at - -
          Rat Genome U34 Set J04486_at - -
          Rat Genome U34 Set M91595exon_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22050716 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360048 - -
          RatRef-12 Expression BeadChip ILMN_54332 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline