Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565482_m1
          See other IL1R1 GE Assays ›
          Gene Symbol:
          Il1r1
          Gene Name:
          interleukin 1 receptor, type I
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 46962292 - 47038139 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565482_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il1r1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013123.3 NP_037255.3 6 - 7 769 63
          XM_006244746.2 XP_006244808.1 7 - 8 2878 63
          XM_006244747.2 XP_006244809.1 7 - 8 1135 63
          XM_006244748.2 XP_006244810.1 6 - 7 1039 63
          XM_006244749.2 XP_006244811.1 8 - 9 958 63
          XM_006244750.2 XP_006244812.1 8 - 9 2950 63
          XM_006244751.2 XP_006244813.1 7 - 8 869 63
          XM_006244752.2 XP_006244814.1 7 - 8 1111 63
          XM_006244753.2 XP_006244815.1 7 - 8 2230 63
          XM_006244754.2 XP_006244816.1 6 - 7 2782 63
          XM_008766993.1 XP_008765215.1 7 - 8 886 63
          GenBank mRNA AB112436.1 - 8 - 9 903 63
          AB112437.1 - 6 - 7 769 63
          AB112438.1 - 7 - 8 841 63
          M95578.1 - 7 - 8 1090 63
          U14010.1 - 5 - 6 718 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000535.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il1r1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to radiation
          heat acclimation
          cytokine-mediated signaling pathway
          ovulation
          regulation of inflammatory response
          interleukin-1-mediated signaling pathway
          response to interleukin-1
          response to epidermal growth factor
          cellular response to glucose stimulus
          response to transforming growth factor beta
          response to nitric oxide
          protease binding
          signal transducer activity
          interleukin-1 receptor activity
          interleukin-1, Type I, activating receptor activity
          platelet-derived growth factor receptor binding
          protein binding
          ionotropic glutamate receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P379588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158232 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175385 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48022 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13808 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P379588 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P379588 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P379588 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P463709 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P379588 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58578 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27054 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10922837 - -
          Affymetrix Rat Gene 1.0 ST Array 10922833 - -
          Affymetrix Rat Gene 1.0 ST Array 10922828 - -
          Affymetrix Rat Gene 1.0 ST Array 10922832 - -
          Affymetrix Rat Gene 1.0 ST Array 10922840 - -
          Affymetrix Rat Gene 1.0 ST Array 10922830 - -
          Affymetrix Rat Gene 1.0 ST Array 10922827 - -
          Affymetrix Rat Gene 1.0 ST Array 10922841 - -
          Affymetrix Rat Gene 1.0 ST Array 10922842 - -
          Affymetrix Rat Gene 1.0 ST Array 10922831 - -
          Affymetrix Rat Gene 1.0 ST Array 10922836 - -
          Affymetrix Rat Gene 1.0 ST Array 10922834 - -
          Affymetrix Rat Gene 1.0 ST Array 10922839 - -
          Affymetrix Rat Gene 1.0 ST Array 10922838 - -
          Affymetrix Rat Gene 1.0 ST Array 10922835 - -
          Affymetrix Rat Gene 1.0 ST Array 10922829 - -
          Affymetrix Rat Gene 1.1 ST Array 10922826 - -
          Rat Genome 230 2.0 Array 1369255_PM_at - -
          Rat Genome 230 2.0 Array 1370750_PM_a_at - -
          Rat Genome 230 2.0 Array 1370750_a_at - -
          Rat Genome 230 2.0 Array 1369255_at - -
          Affymetrix HT Rat Focus Array 1369255_at - -
          Affymetrix HT Rat Focus Array 1370750_a_at - -
          Affymetrix HT Rat Focus Array 1369255_PM_at - -
          Affymetrix HT Rat Focus Array 1370750_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0900004364.rn.1 TC0900000535.rn.1 RTA TC
          Rat Expression Set 230 1370750_a_at - -
          Rat Expression Set 230 1369255_PM_at - -
          Rat Expression Set 230 1369255_at - -
          Rat Expression Set 230 1370750_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10922826 - -
          Rat Genome U34 Set U14010_at - -
          Rat Genome U34 Set M95578_g_at - -
          Rat Genome U34 Set M95578_at - -
          Rat Genome U34 Set U14010_g_at - -
          Rat Genome U34 Set U14010_g_at - -
          Rat Genome U34 Set M95578_g_at - -
          Rat Genome U34 Set U14010_at - -
          Rat Genome U34 Set M95578_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22398781 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356797 - -
          RatRef-12 Expression BeadChip ILMN_56725 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline