Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565571_m1
          See other ANXA5 GE Assays ›
          Gene Symbol:
          Anxa5
          Gene Name:
          annexin A5
          Gene Aliases:
          Anx5, LC5
          Chromosome Location:
          Chr.2: 123162477 - 123193178 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565571_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Anxa5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013132.1 NP_037264.1 4 - 5 252 115
          XM_006232268.2 XP_006232330.1 4 - 5 387 115
          GenBank mRNA AF051895.1 - 4 - 5 307 115
          BC081855.1 - 4 - 5 305 7192977 115
          FQ213209.1 - 5 - 6 382 115
          FQ229127.1 - 4 - 5 240 115
          M21730.1 - 4 - 5 252 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003517.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Anxa5

          Panther Classification:

          Molecular Function -

          calcium-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          blood coagulation
          response to organic substance
          negative regulation of blood coagulation
          positive regulation of apoptotic process
          negative regulation of coagulation
          protein homooligomerization
          response to calcium ion
          regulation of sperm motility
          calcium ion binding
          protein binding
          calcium-dependent phospholipid binding
          peptide hormone binding
          receptor tyrosine kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P472994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130967 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95203 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P472994 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P472994 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P472994 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P472996 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P472994 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700590390 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66037 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9953 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27714 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41178 - -
          Agilent Rat cDNA Microarray (G4105A) 700590390 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10822838 - -
          Affymetrix Rat Gene 1.0 ST Array 10822849 - -
          Affymetrix Rat Gene 1.0 ST Array 10822846 - -
          Affymetrix Rat Gene 1.0 ST Array 10822847 - -
          Affymetrix Rat Gene 1.0 ST Array 10822851 - -
          Affymetrix Rat Gene 1.0 ST Array 10822845 - -
          Affymetrix Rat Gene 1.0 ST Array 10822840 - -
          Affymetrix Rat Gene 1.0 ST Array 10822839 - -
          Affymetrix Rat Gene 1.0 ST Array 10822841 - -
          Affymetrix Rat Gene 1.0 ST Array 10822843 - -
          Affymetrix Rat Gene 1.0 ST Array 10822844 - -
          Affymetrix Rat Gene 1.0 ST Array 10822850 - -
          Affymetrix Rat Gene 1.0 ST Array 10822848 - -
          Affymetrix Rat Gene 1.0 ST Array 10822842 - -
          Affymetrix Rat Gene 1.1 ST Array 10822837 - -
          Rat Genome 230 2.0 Array 1386862_PM_at - -
          Rat Genome 230 2.0 Array 1386862_at - -
          Affymetrix HT Rat Focus Array 1386862_PM_at - -
          Affymetrix HT Rat Focus Array 1386862_at - -
          Affymetrix Rat Clariom S Assay PSR0200023895.rn.1 TC0200003517.rn.1 RTA TC
          Rat Expression Set 230 1386862_PM_at - -
          Rat Expression Set 230 1386862_at - -
          Affymetrix Rat Gene 1.0 ST Array 10822837 - -
          Rat Genome U34 Set D42137exon_s_at - -
          Rat Genome U34 Set AF051895_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22291619 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370963 - -
          RatRef-12 Expression BeadChip ILMN_52928 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0