Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565664_m1
          See other ITPR3 GE Assays ›
          Gene Symbol:
          Itpr3
          Gene Name:
          inositol 1,4,5-trisphosphate receptor, type 3
          Gene Aliases:
          IP3R3, IP3R3X
          Chromosome Location:
          Chr.20: 5645942 - 5711707 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565664_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itpr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013138.1 NP_037270.1 22 - 23 3060 63
          XM_008772710.1 XP_008770932.1 23 - 24 3124 63
          GenBank mRNA GQ233031.1 - 22 - 23 2926 63
          L06096.1 - 22 - 23 3060 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000229.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itpr3

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          memory
          inositol phosphate-mediated signaling
          sensory perception of bitter taste
          sensory perception of sweet taste
          sensory perception of umami taste
          protein homooligomerization
          protein heterooligomerization
          response to calcium ion
          long-term synaptic potentiation
          calcium ion transport into cytosol
          calcium ion transmembrane transport
          cellular response to cAMP
          inositol hexakisphosphate binding
          inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
          calcium ion binding
          calcium ion transmembrane transporter activity
          calcium-release channel activity
          phosphatidylinositol binding
          inositol 1,3,4,5 tetrakisphosphate binding
          inositol 1,4,5 trisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P288185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85324 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163694 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126089 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115283 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288185 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P288185 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P288185 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288187 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288185 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16758 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74746 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94691 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80817 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89577 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55257 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828543 - -
          Affymetrix Rat Gene 1.0 ST Array 10828552 - -
          Affymetrix Rat Gene 1.0 ST Array 10828540 - -
          Affymetrix Rat Gene 1.0 ST Array 10828564 - -
          Affymetrix Rat Gene 1.0 ST Array 10828521 - -
          Affymetrix Rat Gene 1.0 ST Array 10828530 - -
          Affymetrix Rat Gene 1.0 ST Array 10828563 - -
          Affymetrix Rat Gene 1.0 ST Array 10828545 - -
          Affymetrix Rat Gene 1.0 ST Array 10828528 - -
          Affymetrix Rat Gene 1.0 ST Array 10828561 - -
          Affymetrix Rat Gene 1.0 ST Array 10828569 - -
          Affymetrix Rat Gene 1.0 ST Array 10828524 - -
          Affymetrix Rat Gene 1.0 ST Array 10828535 - -
          Affymetrix Rat Gene 1.0 ST Array 10828519 - -
          Affymetrix Rat Gene 1.0 ST Array 10828529 - -
          Affymetrix Rat Gene 1.0 ST Array 10828550 - -
          Affymetrix Rat Gene 1.0 ST Array 10828532 - -
          Affymetrix Rat Gene 1.0 ST Array 10828520 - -
          Affymetrix Rat Gene 1.0 ST Array 10828554 - -
          Affymetrix Rat Gene 1.0 ST Array 10828522 - -
          Affymetrix Rat Gene 1.0 ST Array 10828541 - -
          Affymetrix Rat Gene 1.0 ST Array 10828518 - -
          Affymetrix Rat Gene 1.0 ST Array 10828527 - -
          Affymetrix Rat Gene 1.0 ST Array 10828539 - -
          Affymetrix Rat Gene 1.0 ST Array 10828538 - -
          Affymetrix Rat Gene 1.0 ST Array 10828549 - -
          Affymetrix Rat Gene 1.0 ST Array 10828555 - -
          Affymetrix Rat Gene 1.0 ST Array 10828573 - -
          Affymetrix Rat Gene 1.0 ST Array 10828557 - -
          Affymetrix Rat Gene 1.0 ST Array 10828526 - -
          Affymetrix Rat Gene 1.0 ST Array 10828551 - -
          Affymetrix Rat Gene 1.0 ST Array 10828523 - -
          Affymetrix Rat Gene 1.0 ST Array 10828567 - -
          Affymetrix Rat Gene 1.0 ST Array 10828571 - -
          Affymetrix Rat Gene 1.0 ST Array 10828544 - -
          Affymetrix Rat Gene 1.0 ST Array 10828566 - -
          Affymetrix Rat Gene 1.0 ST Array 10828537 - -
          Affymetrix Rat Gene 1.0 ST Array 10828542 - -
          Affymetrix Rat Gene 1.0 ST Array 10828562 - -
          Affymetrix Rat Gene 1.0 ST Array 10828536 - -
          Affymetrix Rat Gene 1.0 ST Array 10828556 - -
          Affymetrix Rat Gene 1.0 ST Array 10828558 - -
          Affymetrix Rat Gene 1.0 ST Array 10828548 - -
          Affymetrix Rat Gene 1.0 ST Array 10828559 - -
          Affymetrix Rat Gene 1.0 ST Array 10828568 - -
          Affymetrix Rat Gene 1.0 ST Array 10828534 - -
          Affymetrix Rat Gene 1.0 ST Array 10828517 - -
          Affymetrix Rat Gene 1.0 ST Array 10828533 - -
          Affymetrix Rat Gene 1.0 ST Array 10828565 - -
          Affymetrix Rat Gene 1.0 ST Array 10828572 - -
          Affymetrix Rat Gene 1.0 ST Array 10828560 - -
          Affymetrix Rat Gene 1.0 ST Array 10828553 - -
          Affymetrix Rat Gene 1.0 ST Array 10828531 - -
          Affymetrix Rat Gene 1.0 ST Array 10828570 - -
          Affymetrix Rat Gene 1.0 ST Array 10828525 - -
          Affymetrix Rat Gene 1.0 ST Array 10828547 - -
          Affymetrix Rat Gene 1.1 ST Array 10828516 - -
          Rat Genome 230 2.0 Array 1368005_at - -
          Rat Genome 230 2.0 Array 1368005_PM_at - -
          Affymetrix HT Rat Focus Array 1368005_PM_at - -
          Affymetrix HT Rat Focus Array 1368005_at - -
          Affymetrix Rat Clariom S Assay PSR2000002246.rn.1 TC2000000229.rn.1 RTA TC
          Rat Expression Set 230 1368005_at - -
          Rat Expression Set 230 1368005_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828516 - -
          Rat Genome U34 Set L06096_g_at - -
          Rat Genome U34 Set L06096_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21633816 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357844 - -
          RatRef-12 Expression BeadChip ILMN_52597 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline