Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565707_m1
          See other PPARD GE Assays ›
          Gene Symbol:
          Ppard
          Gene Name:
          peroxisome proliferator-activated receptor delta
          Gene Aliases:
          Pparb
          Chromosome Location:
          Chr.20: 7818289 - 7883482 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565707_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (2)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppard
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013141.2 NP_037273.2 3 - 4 360 94
          XM_006256166.2 XP_006256228.1 3 - 4 1172 94
          XM_006256167.2 XP_006256229.1 2 - 3 1279 94
          XM_006256168.2 XP_006256230.1 2 - 3 2174 94
          XM_008772797.1 XP_008771019.1 2 - 3 1407 94
          GenBank mRNA AJ306400.1 - 1 - 2 126 94
          FM033221.1 - 3 - 4 360 94
          U40064.1 - 3 - 4 381 94
          U75918.1 - 3 - 4 374 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000312.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Nuclear Receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppard

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          placenta development
          proteoglycan metabolic process
          lipid metabolic process
          fatty acid beta-oxidation
          vitamin A metabolic process
          heart development
          embryo implantation
          cell proliferation
          positive regulation of cell proliferation
          axon ensheathment
          phospholipid biosynthetic process
          mRNA transcription
          response to glucose
          response to organic substance
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to activity
          negative regulation of smooth muscle cell migration
          fatty acid transport
          fatty acid oxidation
          cell differentiation
          negative regulation of cell growth
          intracellular receptor signaling pathway
          cell-substrate adhesion
          positive regulation of insulin secretion
          negative regulation of collagen biosynthetic process
          response to vitamin A
          wound healing
          regulation of cell proliferation
          negative regulation of apoptotic process
          steroid hormone mediated signaling pathway
          keratinocyte proliferation
          regulation of fat cell differentiation
          positive regulation of epidermis development
          positive regulation of transcription, DNA-templated
          positive regulation of vasodilation
          decidualization
          negative regulation of smooth muscle cell proliferation
          negative regulation of epithelial cell proliferation
          negative regulation of inflammatory response
          keratinocyte migration
          adipose tissue development
          cellular response to hypoxia
          apoptotic signaling pathway
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          transcription coactivator activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          fatty acid binding
          drug binding
          zinc ion binding
          lipid binding
          sequence-specific DNA binding
          protein heterodimerization activity
          NF-kappaB binding
          linoleic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P051712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65156 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30887 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84496 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97281 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174542 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138424 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72438 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P458711 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051711 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051711 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P458711 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P458711 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P051711 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11733 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701899077 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64369 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20454 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51364 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14900 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67721 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42618 - -
          Agilent Rat cDNA Microarray (G4105A) 701899077 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828718 - -
          Affymetrix Rat Gene 1.0 ST Array 10828721 - -
          Affymetrix Rat Gene 1.0 ST Array 10828722 - -
          Affymetrix Rat Gene 1.0 ST Array 10828719 - -
          Affymetrix Rat Gene 1.0 ST Array 10828717 - -
          Affymetrix Rat Gene 1.0 ST Array 10828716 - -
          Affymetrix Rat Gene 1.0 ST Array 10828720 - -
          Affymetrix Rat Gene 1.1 ST Array 10828714 - -
          Rat Genome 230 2.0 Array 1374914_PM_at - -
          Rat Genome 230 2.0 Array 1387684_PM_at - -
          Rat Genome 230 2.0 Array 1387684_at - -
          Rat Genome 230 2.0 Array 1374914_at - -
          Affymetrix HT Rat Focus Array 1374914_PM_at - -
          Affymetrix HT Rat Focus Array 1374914_at - -
          Affymetrix HT Rat Focus Array 1387684_at - -
          Affymetrix HT Rat Focus Array 1387684_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000002912.rn.1 TC2000000312.rn.1 RTA TC
          Rat Expression Set 230 1374914_PM_at - -
          Rat Expression Set 230 1387684_at - -
          Rat Expression Set 230 1374914_at - -
          Rat Expression Set 230 1387684_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828714 - -
          Rat Genome U34 Set rc_AI230294_at - -
          Rat Genome U34 Set U40064_g_at - -
          Rat Genome U34 Set U40064_at - -
          Rat Genome U34 Set U40064_at - -
          Rat Genome U34 Set rc_AI230294_at - -
          Rat Genome U34 Set U40064_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21338089 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357865 - -
          RatRef-12 Expression BeadChip ILMN_53359 - -

          Back To Top
           

          Citations (2)

          Causes and prevention of tamoxifen-induced accumulation of triacylglycerol in rat liver.
          Authors
          Gudbrandsen OA, Rost TH, Berge RK
          Journal
          J Lipid Res - Page No 2223 (Sun Oct 01 00:00:00 EDT 2006)
          Volume (Year)
          47 (2006)
          Product Usage
          -
          Early postnatal oestradiol exposure causes insulin resistance and signs of inflammation in circulation and skeletal muscle.
          Authors
          Alexanderson C, Eriksson E, Stener-Victorin E, Lonn M, Holmang A
          Journal
          J Endocrinol - Page No 49 (Wed Apr 01 00:00:00 EDT 2009)
          Volume (Year)
          201 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline