Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565736_m1
          See other CLCN1 GE Assays ›
          Gene Symbol:
          Clcn1
          Gene Name:
          chloride channel, voltage-sensitive 1
          Gene Aliases:
          SMCC
          Chromosome Location:
          Chr.4: 71674467 - 71704159 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565736_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clcn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013147.1 NP_037279.1 20 - 21 2510 133
          XM_008762873.1 XP_008761095.1 19 - 20 133
          GenBank mRNA AY112736.1 - 19 - 20 2259 133
          AY112737.1 - 21 - 22 2420 133
          AY112738.1 - 20 - 21 2341 133
          AY112739.1 - 19 - 20 2212 133
          AY112740.1 - 20 - 21 2338 133
          X62894.1 - 20 - 21 2510 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000754.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clcn1

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          chloride transport
          neuronal action potential propagation
          ion transmembrane transport
          regulation of ion transmembrane transport
          regulation of anion transport
          chloride transmembrane transport
          voltage-gated chloride channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P018102 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63238 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174024 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27901 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018102 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11736 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018102 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11736 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P018102 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11736 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P529311 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2808 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90478 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855060 - -
          Affymetrix Rat Gene 1.0 ST Array 10855036 - -
          Affymetrix Rat Gene 1.0 ST Array 10855042 - -
          Affymetrix Rat Gene 1.0 ST Array 10855057 - -
          Affymetrix Rat Gene 1.0 ST Array 10855045 - -
          Affymetrix Rat Gene 1.0 ST Array 10855039 - -
          Affymetrix Rat Gene 1.0 ST Array 10855040 - -
          Affymetrix Rat Gene 1.0 ST Array 10855046 - -
          Affymetrix Rat Gene 1.0 ST Array 10855049 - -
          Affymetrix Rat Gene 1.0 ST Array 10855059 - -
          Affymetrix Rat Gene 1.0 ST Array 10855061 - -
          Affymetrix Rat Gene 1.0 ST Array 10855041 - -
          Affymetrix Rat Gene 1.0 ST Array 10855038 - -
          Affymetrix Rat Gene 1.0 ST Array 10855037 - -
          Affymetrix Rat Gene 1.0 ST Array 10855055 - -
          Affymetrix Rat Gene 1.0 ST Array 10855050 - -
          Affymetrix Rat Gene 1.0 ST Array 10855047 - -
          Affymetrix Rat Gene 1.0 ST Array 10855053 - -
          Affymetrix Rat Gene 1.0 ST Array 10855043 - -
          Affymetrix Rat Gene 1.0 ST Array 10855044 - -
          Affymetrix Rat Gene 1.0 ST Array 10855056 - -
          Affymetrix Rat Gene 1.0 ST Array 10855058 - -
          Affymetrix Rat Gene 1.0 ST Array 10855054 - -
          Affymetrix Rat Gene 1.0 ST Array 10855048 - -
          Affymetrix Rat Gene 1.0 ST Array 10855052 - -
          Affymetrix Rat Gene 1.1 ST Array 10855035 - -
          Rat Genome 230 2.0 Array 1387719_at - -
          Rat Genome 230 2.0 Array 1387719_PM_at - -
          Affymetrix HT Rat Focus Array 1387719_at - -
          Affymetrix HT Rat Focus Array 1387719_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400005567.rn.1 TC0400000754.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400005568.rn.1 TC0400000754.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400005569.rn.1 TC0400000754.rn.1 RTA TC
          Rat Expression Set 230 1387719_at - -
          Rat Expression Set 230 1387719_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855035 - -
          Rat Genome U34 Set X62894_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21142472 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362416 - -
          RatRef-12 Expression BeadChip ILMN_68485 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline