Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565750_m1
          See other AHR GE Assays ›
          Gene Symbol:
          Ahr
          Gene Name:
          aryl hydrocarbon receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 54966533 - 55001436 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565750_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ahr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013149.2 NP_037281.2 3 - 4 350 62
          GenBank mRNA AF082124.1 - 3 - 4 379 62
          AF082125.1 - 3 - 4 379 62
          AF082126.1 - 3 - 4 379 62
          AJ821851.1 - 3 - 4 323 62
          AM902281.1 - 3 - 4 350 62
          AM902282.1 - 3 - 4 350 62
          AM902283.1 - 3 - 4 350 62
          AM902284.1 - 3 - 4 350 62
          AM902285.1 - 3 - 4 350 62
          AM902286.1 - 3 - 4 350 62
          U09000.1 - 3 - 4 350 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002326.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ahr

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          ovarian follicle development
          blood vessel development
          patterning of blood vessels
          B cell homeostasis
          liver development
          blood vessel remodeling
          lymphocyte homeostasis
          immune system process
          negative regulation of systemic arterial blood pressure
          cardiac left ventricle morphogenesis
          circumferential growth involved in left ventricle morphogenesis
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          apoptotic process
          blood circulation
          response to xenobiotic stimulus
          response to toxic substance
          regulation of gene expression
          response to organic cyclic compound
          intracellular receptor signaling pathway
          prostate gland development
          regulation of B cell proliferation
          response to estradiol
          circadian regulation of gene expression
          embryonic hemopoiesis
          post-embryonic hemopoiesis
          camera-type eye development
          T cell homeostasis
          positive regulation of cell size
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
          negative regulation of vasoconstriction
          positive regulation of growth
          positive regulation of transcription from RNA polymerase II promoter
          blood vessel morphogenesis
          spleen development
          reproductive structure development
          gland development
          smooth muscle tissue development
          regulation of blood vessel size
          negative regulation of necrotic cell death
          venous blood vessel development
          kidney morphogenesis
          common bile duct development
          cellular response to cAMP
          cellular response to glucocorticoid stimulus
          glomerulus morphogenesis
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          signal transducer activity
          aryl hydrocarbon receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          transcription factor binding
          enhancer binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity
          protein dimerization activity
          Hsp90 protein binding
          E-box binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0600016794.rn.1 TC0600002326.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600016793.rn.1 TC0600002326.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline