Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565819_m1
          See other DBH GE Assays ›
          Gene Symbol:
          Dbh
          Gene Name:
          dopamine beta-hydroxylase (dopamine beta-monooxygenase)
          Gene Aliases:
          DOPBHY
          Chromosome Location:
          Chr.3: 5709236 - 5731895 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565819_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dbh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013158.2 NP_037290.2 6 - 7 1209 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000137.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dbh

          Panther Classification:

          Molecular Function -

          hydroxylase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production
          blood vessel remodeling
          response to amphetamine
          leukocyte mediated immunity
          catecholamine metabolic process
          memory
          locomotory behavior
          associative learning
          visual learning
          response to iron ion
          response to ozone
          multicellular organism aging
          response to organic cyclic compound
          response to estradiol
          social behavior
          response to isolation stress
          response to immobilization stress
          regulation of cell proliferation
          homoiothermy
          dopamine catabolic process
          norepinephrine biosynthetic process
          response to drug
          glucose homeostasis
          fear response
          maternal behavior
          response to peptide hormone
          response to ethanol
          positive regulation of vasoconstriction
          octopamine metabolic process
          response to copper ion
          behavioral response to ethanol
          response to pain
          leukocyte migration
          sensory perception of taste
          oxidation-reduction process
          bone development
          cellular response to manganese ion
          cellular response to nicotine
          regulation of extrinsic apoptotic signaling pathway
          dopamine beta-monooxygenase activity
          copper ion binding
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced ascorbate as one donor, and incorporation of one atom of oxygen
          L-ascorbic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P083610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260903 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53259 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130974 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P083610 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P083610 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P083610 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P381686 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31725 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10834694 - -
          Affymetrix Rat Gene 1.0 ST Array 10834702 - -
          Affymetrix Rat Gene 1.0 ST Array 10834701 - -
          Affymetrix Rat Gene 1.0 ST Array 10834696 - -
          Affymetrix Rat Gene 1.0 ST Array 10834699 - -
          Affymetrix Rat Gene 1.0 ST Array 10834695 - -
          Affymetrix Rat Gene 1.0 ST Array 10834697 - -
          Affymetrix Rat Gene 1.0 ST Array 10834700 - -
          Affymetrix Rat Gene 1.0 ST Array 10834703 - -
          Affymetrix Rat Gene 1.0 ST Array 10834698 - -
          Affymetrix Rat Gene 1.0 ST Array 10834693 - -
          Affymetrix Rat Gene 1.0 ST Array 10834692 - -
          Affymetrix Rat Gene 1.1 ST Array 10834691 - -
          Rat Genome 230 2.0 Array 1370564_at - -
          Rat Genome 230 2.0 Array 1370564_PM_at - -
          Affymetrix HT Rat Focus Array 1370564_PM_at - -
          Affymetrix HT Rat Focus Array 1370564_at - -
          Affymetrix Rat Clariom S Assay PSR0300001310.rn.1 TC0300000137.rn.1 RTA TC
          Rat Expression Set 230 1370564_at - -
          Rat Expression Set 230 1370564_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10834691 - -
          Rat Genome U34 Set L12407_at - -
          Rat Genome U34 Set L12407_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21731357 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374582 - -
          RatRef-12 Expression BeadChip ILMN_66537 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline