Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565867_m1
          See other CCKBR GE Assays ›
          Gene Symbol:
          Cckbr
          Gene Name:
          cholecystokinin B receptor
          Gene Aliases:
          Cck2r, Cholrec
          Chromosome Location:
          Chr.1: 170262218 - 170272298 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565867_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cckbr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013165.2 NP_037297.1 1 - 2 194 109
          GenBank mRNA M99418.1 - 1 - 2 285 109
          X79208.1 - 1 - 2 150 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002279.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cckbr

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          gastric acid secretion
          histamine secretion
          behavioral defense response
          apoptotic process
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          digestion
          positive regulation of cell proliferation
          positive regulation of synaptic transmission, GABAergic
          response to insulin
          estrous cycle
          digestive tract development
          gland development
          positive regulation of synaptic transmission
          regulation of sensory perception of pain
          positive regulation of synaptic transmission, glutamatergic
          ERK1 and ERK2 cascade
          positive regulation of somatostatin secretion
          gastrin receptor activity
          type B gastrin/cholecystokinin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P101130 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P013932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535133 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156049 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101130 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013932 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013932 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101130 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P013932 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P101130 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P556083 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7109 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10709567 - -
          Affymetrix Rat Gene 1.0 ST Array 10709574 - -
          Affymetrix Rat Gene 1.0 ST Array 10709571 - -
          Affymetrix Rat Gene 1.0 ST Array 10709566 - -
          Affymetrix Rat Gene 1.0 ST Array 10709573 - -
          Affymetrix Rat Gene 1.0 ST Array 10709569 - -
          Affymetrix Rat Gene 1.0 ST Array 10709572 - -
          Affymetrix Rat Gene 1.0 ST Array 10709570 - -
          Affymetrix Rat Gene 1.0 ST Array 10709568 - -
          Affymetrix Rat Gene 1.1 ST Array 10709565 - -
          Rat Genome 230 2.0 Array 1371159_PM_a_at - -
          Rat Genome 230 2.0 Array 1369713_PM_at - -
          Rat Genome 230 2.0 Array 1369713_at - -
          Rat Genome 230 2.0 Array 1371159_a_at - -
          Affymetrix HT Rat Focus Array 1369713_PM_at - -
          Affymetrix HT Rat Focus Array 1371159_a_at - -
          Affymetrix HT Rat Focus Array 1371159_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369713_at - -
          Affymetrix Rat Clariom S Assay PSR0100017364.rn.1 TC0100002279.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100017365.rn.1 TC0100002279.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100017367.rn.1 TC0100002279.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100017366.rn.1 TC0100002279.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100017368.rn.1 TC0100002279.rn.1 RTA TC
          Rat Expression Set 230 1371159_a_at - -
          Rat Expression Set 230 1369713_at - -
          Rat Expression Set 230 1371159_PM_a_at - -
          Rat Expression Set 230 1369713_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10709565 - -
          Rat Genome U34 Set X79208_s_at - -
          Rat Genome U34 Set M99418_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21172219 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351032 - -
          RatRef-12 Expression BeadChip ILMN_52411 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline