Search
Search
| Gene Symbol: Tgfb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013174.2 | NP_037306.1 | 3 - 4 | 1050 | 61 | |
| XM_006240366.1 | XP_006240428.1 | 4 - 5 | 1394 | 61 | ||
| GenBank mRNA | BC092195.1 | - | 3 - 4 | 1037 | 7313608 | 61 |
| U03491.1 | - | 3 - 4 | 1050 | 61 | ||
Set Membership: |
|