Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566017_m1
          See other ITGB4 GE Assays ›
          Gene Symbol:
          Itgb4
          Gene Name:
          integrin, beta 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 104524000 - 104560180 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566017_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itgb4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013180.1 NP_037312.1 30 - 31 4001 66
          GenBank mRNA U60096.1 - 30 - 31 4001 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001954.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itgb4

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          multicellular organism development
          response to wounding
          hemidesmosome assembly
          nail development
          skin development
          filopodium assembly
          mesodermal cell differentiation
          digestive tract development
          cell motility
          renal system development
          amelogenesis
          G-protein coupled receptor binding
          receptor activity
          cell adhesion molecule binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P055738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141005 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105926 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85767 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055738 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055738 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P055738 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P490296 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700228850 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12162 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63980 - -
          Agilent Rat cDNA Microarray (G4105A) 700228850 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10739748 - -
          Affymetrix Rat Gene 1.0 ST Array 10739757 - -
          Affymetrix Rat Gene 1.0 ST Array 10739744 - -
          Affymetrix Rat Gene 1.0 ST Array 10739749 - -
          Affymetrix Rat Gene 1.0 ST Array 10739727 - -
          Affymetrix Rat Gene 1.0 ST Array 10739737 - -
          Affymetrix Rat Gene 1.0 ST Array 10739736 - -
          Affymetrix Rat Gene 1.0 ST Array 10739746 - -
          Affymetrix Rat Gene 1.0 ST Array 10739747 - -
          Affymetrix Rat Gene 1.0 ST Array 10739733 - -
          Affymetrix Rat Gene 1.0 ST Array 10739760 - -
          Affymetrix Rat Gene 1.0 ST Array 10739750 - -
          Affymetrix Rat Gene 1.0 ST Array 10739762 - -
          Affymetrix Rat Gene 1.0 ST Array 10739723 - -
          Affymetrix Rat Gene 1.0 ST Array 10739742 - -
          Affymetrix Rat Gene 1.0 ST Array 10739740 - -
          Affymetrix Rat Gene 1.0 ST Array 10739739 - -
          Affymetrix Rat Gene 1.0 ST Array 10739738 - -
          Affymetrix Rat Gene 1.0 ST Array 10739735 - -
          Affymetrix Rat Gene 1.0 ST Array 10739731 - -
          Affymetrix Rat Gene 1.0 ST Array 10739753 - -
          Affymetrix Rat Gene 1.0 ST Array 10739729 - -
          Affymetrix Rat Gene 1.0 ST Array 10739759 - -
          Affymetrix Rat Gene 1.0 ST Array 10739755 - -
          Affymetrix Rat Gene 1.0 ST Array 10739732 - -
          Affymetrix Rat Gene 1.0 ST Array 10739730 - -
          Affymetrix Rat Gene 1.0 ST Array 10739756 - -
          Affymetrix Rat Gene 1.0 ST Array 10739751 - -
          Affymetrix Rat Gene 1.0 ST Array 10739745 - -
          Affymetrix Rat Gene 1.0 ST Array 10739734 - -
          Affymetrix Rat Gene 1.0 ST Array 10739763 - -
          Affymetrix Rat Gene 1.0 ST Array 10739728 - -
          Affymetrix Rat Gene 1.0 ST Array 10739741 - -
          Affymetrix Rat Gene 1.0 ST Array 10739722 - -
          Affymetrix Rat Gene 1.0 ST Array 10739724 - -
          Affymetrix Rat Gene 1.0 ST Array 10739758 - -
          Affymetrix Rat Gene 1.0 ST Array 10739761 - -
          Affymetrix Rat Gene 1.0 ST Array 10739752 - -
          Affymetrix Rat Gene 1.0 ST Array 10739726 - -
          Affymetrix Rat Gene 1.1 ST Array 10739721 - -
          Rat Genome 230 2.0 Array 1368612_at - -
          Rat Genome 230 2.0 Array 1368612_PM_at - -
          Affymetrix HT Rat Focus Array 1368612_at - -
          Affymetrix HT Rat Focus Array 1368612_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000017655.rn.1 TC1000001954.rn.1 RTA TC
          Rat Expression Set 230 1368612_at - -
          Rat Expression Set 230 1368612_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10739721 - -
          Rat Genome U34 Set U60096_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21959171 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374720 - -
          RatRef-12 Expression BeadChip ILMN_59068 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0