Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566082_m1
          See other HCK GE Assays ›
          Gene Symbol:
          Hck
          Gene Name:
          hemopoietic cell kinase
          Gene Aliases:
          Hctk, p56Hck, p59Hck
          Chromosome Location:
          Chr.3: 148579917 - 148623026 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566082_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hck
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013185.3 NP_037317.2 2 - 3 378 76
          GenBank mRNA BC078890.1 - 2 - 3 378 7121407 76
          M83666.1 - 2 - 3 300 76
          S74141.1 - 2 - 3 300 76
          X62345.1 - 2 - 3 329 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002057.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hck

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          phagocytosis
          inflammatory response
          cytoskeleton organization
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of cell proliferation
          regulation of cell shape
          peptidyl-tyrosine phosphorylation
          cell differentiation
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          negative regulation of apoptotic process
          innate immune response
          protein autophosphorylation
          regulation of phagocytosis
          defense response to Gram-positive bacterium
          regulation of sequence-specific DNA binding transcription factor activity
          cellular response to peptide hormone stimulus
          regulation of podosome assembly
          positive regulation of actin cytoskeleton reorganization
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528977 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161417 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109956 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114438 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105162 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11749 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11749 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11749 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11749 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700482506 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45803 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100696 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103794 - -
          Agilent Rat cDNA Microarray (G4105A) 700482506 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840988 - -
          Affymetrix Rat Gene 1.0 ST Array 10840978 - -
          Affymetrix Rat Gene 1.0 ST Array 10840987 - -
          Affymetrix Rat Gene 1.0 ST Array 10840983 - -
          Affymetrix Rat Gene 1.0 ST Array 10840984 - -
          Affymetrix Rat Gene 1.0 ST Array 10840977 - -
          Affymetrix Rat Gene 1.0 ST Array 10840976 - -
          Affymetrix Rat Gene 1.0 ST Array 10840986 - -
          Affymetrix Rat Gene 1.0 ST Array 10840981 - -
          Affymetrix Rat Gene 1.0 ST Array 10840980 - -
          Affymetrix Rat Gene 1.0 ST Array 10840979 - -
          Affymetrix Rat Gene 1.0 ST Array 10840985 - -
          Affymetrix Rat Gene 1.0 ST Array 10840989 - -
          Affymetrix Rat Gene 1.0 ST Array 10840982 - -
          Affymetrix Rat Gene 1.1 ST Array 10840975 - -
          Rat Genome 230 2.0 Array 1369204_at - -
          Rat Genome 230 2.0 Array 1369204_PM_at - -
          Affymetrix HT Rat Focus Array 1369204_at - -
          Affymetrix HT Rat Focus Array 1369204_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300014766.rn.1 TC0300002057.rn.1 RTA TC
          Rat Expression Set 230 1369204_PM_at - -
          Rat Expression Set 230 1369204_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840975 - -
          Rat Genome U34 Set S74141_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21274110 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373013 - -
          RatRef-12 Expression BeadChip ILMN_69202 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline