Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566108_m1
          See other PLCG1 GE Assays ›
          Gene Symbol:
          Plcg1
          Gene Name:
          phospholipase C, gamma 1
          Gene Aliases:
          PPLCA
          Chromosome Location:
          Chr.3: 156727642 - 156758307 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566108_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plcg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013187.1 NP_037319.1 27 - 28 3373 63
          GenBank mRNA J03806.1 - 27 - 28 3373 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002217.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plcg1

          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          calcium ion transport
          signal transduction
          epidermal growth factor receptor signaling pathway
          protein secretion
          phospholipid catabolic process
          response to gravity
          response to organonitrogen compound
          positive regulation of epithelial cell migration
          cell migration
          calcium-mediated signaling
          inositol trisphosphate biosynthetic process
          intracellular signal transduction
          response to hydrogen peroxide
          response to morphine
          positive regulation of blood vessel endothelial cell migration
          positive regulation of angiogenesis
          T cell receptor signaling pathway
          positive regulation of release of sequestered calcium ion into cytosol
          cellular response to epidermal growth factor stimulus
          regulation of store-operated calcium channel activity
          phosphatidylinositol phospholipase C activity
          signal transducer activity
          insulin receptor binding
          neurotrophin TRKA receptor binding
          calcium ion binding
          protein binding
          protein kinase binding
          receptor tyrosine kinase binding
          glutamate receptor binding
          phosphoprotein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047047 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P342166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98190 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157468 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136101 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178583 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047047 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P342166 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P342166 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047047 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047047 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P342166 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P342166 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700492145 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700784125 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2994 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60019 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4415 - -
          Agilent Rat cDNA Microarray (G4105A) 700492145 - -
          Agilent Rat cDNA Microarray (G4105A) 700784125 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841870 - -
          Affymetrix Rat Gene 1.0 ST Array 10841861 - -
          Affymetrix Rat Gene 1.0 ST Array 10841865 - -
          Affymetrix Rat Gene 1.0 ST Array 10841856 - -
          Affymetrix Rat Gene 1.0 ST Array 10841886 - -
          Affymetrix Rat Gene 1.0 ST Array 10841878 - -
          Affymetrix Rat Gene 1.0 ST Array 10841854 - -
          Affymetrix Rat Gene 1.0 ST Array 10841852 - -
          Affymetrix Rat Gene 1.0 ST Array 10841859 - -
          Affymetrix Rat Gene 1.0 ST Array 10841858 - -
          Affymetrix Rat Gene 1.0 ST Array 10841867 - -
          Affymetrix Rat Gene 1.0 ST Array 10841881 - -
          Affymetrix Rat Gene 1.0 ST Array 10841871 - -
          Affymetrix Rat Gene 1.0 ST Array 10841872 - -
          Affymetrix Rat Gene 1.0 ST Array 10841873 - -
          Affymetrix Rat Gene 1.0 ST Array 10841855 - -
          Affymetrix Rat Gene 1.0 ST Array 10841876 - -
          Affymetrix Rat Gene 1.0 ST Array 10841883 - -
          Affymetrix Rat Gene 1.0 ST Array 10841857 - -
          Affymetrix Rat Gene 1.0 ST Array 10841864 - -
          Affymetrix Rat Gene 1.0 ST Array 10841877 - -
          Affymetrix Rat Gene 1.0 ST Array 10841879 - -
          Affymetrix Rat Gene 1.0 ST Array 10841882 - -
          Affymetrix Rat Gene 1.0 ST Array 10841869 - -
          Affymetrix Rat Gene 1.0 ST Array 10841863 - -
          Affymetrix Rat Gene 1.0 ST Array 10841868 - -
          Affymetrix Rat Gene 1.0 ST Array 10841880 - -
          Affymetrix Rat Gene 1.0 ST Array 10841875 - -
          Affymetrix Rat Gene 1.0 ST Array 10841874 - -
          Affymetrix Rat Gene 1.0 ST Array 10841885 - -
          Affymetrix Rat Gene 1.0 ST Array 10841884 - -
          Affymetrix Rat Gene 1.0 ST Array 10841851 - -
          Affymetrix Rat Gene 1.0 ST Array 10841860 - -
          Affymetrix Rat Gene 1.0 ST Array 10841862 - -
          Affymetrix Rat Gene 1.1 ST Array 10841850 - -
          Rat Genome 230 2.0 Array 1368183_at - -
          Rat Genome 230 2.0 Array 1368183_PM_at - -
          Affymetrix HT Rat Focus Array 1368183_at - -
          Affymetrix HT Rat Focus Array 1368183_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300016509.rn.1 TC0300002217.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300016510.rn.1 TC0300002217.rn.1 RTA TC
          Rat Expression Set 230 1368183_PM_at - -
          Rat Expression Set 230 1368183_at - -
          Affymetrix Rat Gene 1.0 ST Array 10841850 - -
          Rat Genome U34 Set J03806_at - -
          Rat Genome U34 Set rc_AA943027_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21061023 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368112 - -
          RatRef-12 Expression BeadChip ILMN_66632 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline