Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566428_m1
          See other ACP2 GE Assays ›
          Gene Symbol:
          Acp2
          Gene Name:
          acid phosphatase 2, lysosomal
          Gene Aliases:
          LAP
          Chromosome Location:
          Chr.3: 80020347 - 80030365 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566428_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016988.2 NP_058684.2 3 - 4 331 71
          XM_006234500.1 XP_006234562.1 4 - 5 1253 71
          GenBank mRNA BC081823.1 - 3 - 4 302 7107570 71
          CB720349.1 - 3 - 4 331 71
          M27893.1 - 3 - 4 300 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001130.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acp2

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          lysosome organization
          chemical synaptic transmission
          response to organic substance
          dephosphorylation
          autophagic cell death
          phosphotyrosine binding
          acid phosphatase activity
          phosphoprotein phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P271769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43879 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126893 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P271769 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P271769 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P271769 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P271769 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700530992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51921 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60162 - -
          Agilent Rat cDNA Microarray (G4105A) 700530992 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10837833 - -
          Affymetrix Rat Gene 1.0 ST Array 10837844 - -
          Affymetrix Rat Gene 1.0 ST Array 10837839 - -
          Affymetrix Rat Gene 1.0 ST Array 10837840 - -
          Affymetrix Rat Gene 1.0 ST Array 10837838 - -
          Affymetrix Rat Gene 1.0 ST Array 10837842 - -
          Affymetrix Rat Gene 1.0 ST Array 10837841 - -
          Affymetrix Rat Gene 1.0 ST Array 10837836 - -
          Affymetrix Rat Gene 1.0 ST Array 10837835 - -
          Affymetrix Rat Gene 1.0 ST Array 10837837 - -
          Affymetrix Rat Gene 1.0 ST Array 10837843 - -
          Affymetrix Rat Gene 1.0 ST Array 10837834 - -
          Affymetrix Rat Gene 1.1 ST Array 10837832 - -
          Rat Genome 230 2.0 Array 1367906_at - -
          Rat Genome 230 2.0 Array 1367906_PM_at - -
          Affymetrix HT Rat Focus Array 1367906_PM_at - -
          Affymetrix HT Rat Focus Array 1367906_at - -
          Affymetrix Rat Clariom S Assay PSR0300007935.rn.1 TC0300001130.rn.1 RTA TC
          Rat Expression Set 230 1367906_PM_at - -
          Rat Expression Set 230 1367906_at - -
          Affymetrix Rat Gene 1.0 ST Array 10837832 - -
          Rat Genome U34 Set rc_AI234950_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21277793 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374963 - -
          RatRef-12 Expression BeadChip ILMN_65366 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline