Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566489_m1
          See other C4BPB GE Assays ›
          Gene Symbol:
          C4bpb
          Gene Name:
          complement component 4 binding protein, beta
          Gene Aliases:
          C4bp-ps1
          Chromosome Location:
          Chr.13: 47387173 - 47398113 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566489_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C4bpb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016995.2 NP_058691.2 4 - 5 782 106
          XM_006249705.2 XP_006249767.1 5 - 6 838 106
          XM_008769423.1 XP_008767645.1 5 - 6 655 106
          GenBank mRNA BC088152.1 - 4 - 5 782 7322205 106
          FQ209636.1 - 4 - 5 793 106
          Z50052.1 - 4 - 5 761 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001363.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C4bpb

          Gene Ontology Categories:

          Function(s)

          complement activation, classical pathway
          innate immune response
          positive regulation of protein catabolic process
          negative regulation of complement activation, classical pathway

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143964 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119392 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007633 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007633 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P774912 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701241743 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34144 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65084 - -
          Agilent Rat cDNA Microarray (G4105A) 701241743 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767438 - -
          Affymetrix Rat Gene 1.0 ST Array 10767440 - -
          Affymetrix Rat Gene 1.0 ST Array 10767439 - -
          Affymetrix Rat Gene 1.0 ST Array 10767443 - -
          Affymetrix Rat Gene 1.0 ST Array 10767437 - -
          Affymetrix Rat Gene 1.0 ST Array 10767441 - -
          Affymetrix Rat Gene 1.0 ST Array 10767442 - -
          Affymetrix Rat Gene 1.1 ST Array 10767436 - -
          Rat Genome 230 2.0 Array 1368695_PM_at - -
          Rat Genome 230 2.0 Array 1368695_at - -
          Affymetrix HT Rat Focus Array 1368695_at - -
          Affymetrix HT Rat Focus Array 1368695_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300008984.rn.1 TC1300001363.rn.1 RTA TC
          Rat Expression Set 230 1368695_PM_at - -
          Rat Expression Set 230 1368695_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767436 - -
          Rat Genome U34 Set Z50052_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21151174 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363829 - -
          RatRef-12 Expression BeadChip ILMN_49704 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline