Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566496_m1
          See other CASR GE Assays ›
          Gene Symbol:
          Casr
          Gene Name:
          calcium-sensing receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 67188630 - 67258721 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566496_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Casr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016996.1 NP_058692.1 6 - 7 2305 64
          GenBank mRNA AY214122.1 - 6 - 7 2295 64
          U10354.1 - 6 - 7 2305 64
          U20289.1 - 5 - 6 1931 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000678.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Casr

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          response to ischemia
          calcium ion transport
          cellular calcium ion homeostasis
          apoptotic process
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          JNK cascade
          positive regulation of cell proliferation
          response to metal ion
          positive regulation of gene expression
          response to organic cyclic compound
          positive regulation of insulin secretion
          positive regulation of ATPase activity
          cellular response to hepatocyte growth factor stimulus
          vasodilation
          positive regulation of vasoconstriction
          branching morphogenesis of an epithelial tube
          positive regulation of positive chemotaxis
          response to calcium ion
          regulation of calcium ion transport
          fat pad development
          calcium ion import
          cellular response to vitamin D
          cellular response to glucose stimulus
          cellular response to low-density lipoprotein particle stimulus
          cellular response to hypoxia
          response to fibroblast growth factor
          positive regulation of calcium ion import
          cellular response to peptide
          magnesium ion binding
          G-protein coupled receptor activity
          integrin binding
          calcium ion binding
          drug binding
          polyamine binding
          protein kinase binding
          ion channel binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P118532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138491 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117550 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78869 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127154 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118532 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118532 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P118532 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11765 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701718252 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66105 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94685 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33517 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38805 - -
          Agilent Rat cDNA Microarray (G4105A) 701718252 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754378 - -
          Affymetrix Rat Gene 1.0 ST Array 10754376 - -
          Affymetrix Rat Gene 1.0 ST Array 10754373 - -
          Affymetrix Rat Gene 1.0 ST Array 10754374 - -
          Affymetrix Rat Gene 1.0 ST Array 10754372 - -
          Affymetrix Rat Gene 1.0 ST Array 10754375 - -
          Affymetrix Rat Gene 1.0 ST Array 10754381 - -
          Affymetrix Rat Gene 1.0 ST Array 10754379 - -
          Affymetrix Rat Gene 1.0 ST Array 10754380 - -
          Affymetrix Rat Gene 1.0 ST Array 10754377 - -
          Affymetrix Rat Gene 1.0 ST Array 10754383 - -
          Affymetrix Rat Gene 1.1 ST Array 10754371 - -
          Rat Genome 230 2.0 Array 1369158_PM_at - -
          Rat Genome 230 2.0 Array 1369158_at - -
          Affymetrix HT Rat Focus Array 1369158_PM_at - -
          Affymetrix HT Rat Focus Array 1369158_at - -
          Affymetrix Rat Clariom S Assay PSR1100003681.rn.1 TC1100000678.rn.1 RTA TC
          Rat Expression Set 230 1369158_PM_at - -
          Rat Expression Set 230 1369158_at - -
          Affymetrix Rat Gene 1.0 ST Array 10754371 - -
          Rat Genome U34 Set U10354_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21280449 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040553 - -
          RatRef-12 Expression BeadChip ILMN_55342 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline