Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566561_m1
          See other ERBB2 GE Assays ›
          Gene Symbol:
          Erbb2
          Gene Name:
          v-erb-b2 avian erythroblastic leukemia viral oncogene 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 86367596 - 86391728 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566561_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Erbb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017003.2 NP_058699.2 7 - 8 1019 74
          GenBank mRNA AY116182.1 - 7 - 8 1019 74
          BC061863.1 - 7 - 8 1091 5597835 74
          X03362.1 - 7 - 8 930 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001575.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Hypoxia Panel Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Erbb2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          signal transduction
          cell surface receptor signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          nervous system development
          central nervous system development
          peripheral nervous system development
          heart development
          skeletal muscle tissue development
          neuromuscular junction development
          motor neuron axon guidance
          positive regulation of cell proliferation
          glial cell differentiation
          phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          signal transduction by protein phosphorylation
          positive regulation of cell growth
          response to progesterone
          regulation of microtubule-based process
          negative regulation of immature T cell proliferation in thymus
          wound healing
          regulation of cell proliferation
          response to drug
          myelination
          negative regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          tongue development
          estrous cycle
          regulation of cell differentiation
          positive regulation of translation
          positive regulation of cell adhesion
          positive regulation of transcription from RNA polymerase I promoter
          positive regulation of transcription from RNA polymerase III promoter
          positive regulation of Ras protein signal transduction
          protein autophosphorylation
          sympathetic nervous system development
          response to axon injury
          oligodendrocyte differentiation
          positive regulation of epithelial cell proliferation
          mammary gland involution
          regulation of ERK1 and ERK2 cascade
          RNA polymerase I core binding
          glycoprotein binding
          DNA binding
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          transmembrane signaling receptor activity
          protein binding
          ATP binding
          protein C-terminus binding
          growth factor binding
          protein phosphatase binding
          ubiquitin protein ligase binding
          identical protein binding
          protein heterodimerization activity
          Hsp90 protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109493 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038812 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854837 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157691 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109493 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109493 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038812 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038812 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038817 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109493 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P118540 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103499 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92169 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10737977 - -
          Affymetrix Rat Gene 1.0 ST Array 10737974 - -
          Affymetrix Rat Gene 1.0 ST Array 10737985 - -
          Affymetrix Rat Gene 1.0 ST Array 10737973 - -
          Affymetrix Rat Gene 1.0 ST Array 10737967 - -
          Affymetrix Rat Gene 1.0 ST Array 10737989 - -
          Affymetrix Rat Gene 1.0 ST Array 10737987 - -
          Affymetrix Rat Gene 1.0 ST Array 10737983 - -
          Affymetrix Rat Gene 1.0 ST Array 10737978 - -
          Affymetrix Rat Gene 1.0 ST Array 10737965 - -
          Affymetrix Rat Gene 1.0 ST Array 10737992 - -
          Affymetrix Rat Gene 1.0 ST Array 10737982 - -
          Affymetrix Rat Gene 1.0 ST Array 10737976 - -
          Affymetrix Rat Gene 1.0 ST Array 10737968 - -
          Affymetrix Rat Gene 1.0 ST Array 10737975 - -
          Affymetrix Rat Gene 1.0 ST Array 10737988 - -
          Affymetrix Rat Gene 1.0 ST Array 10737990 - -
          Affymetrix Rat Gene 1.0 ST Array 10737979 - -
          Affymetrix Rat Gene 1.0 ST Array 10737986 - -
          Affymetrix Rat Gene 1.0 ST Array 10737969 - -
          Affymetrix Rat Gene 1.0 ST Array 10737984 - -
          Affymetrix Rat Gene 1.0 ST Array 10737966 - -
          Affymetrix Rat Gene 1.0 ST Array 10737981 - -
          Affymetrix Rat Gene 1.0 ST Array 10737964 - -
          Affymetrix Rat Gene 1.0 ST Array 10737971 - -
          Affymetrix Rat Gene 1.0 ST Array 10737963 - -
          Affymetrix Rat Gene 1.0 ST Array 10737991 - -
          Affymetrix Rat Gene 1.0 ST Array 10737970 - -
          Affymetrix Rat Gene 1.0 ST Array 10737980 - -
          Affymetrix Rat Gene 1.1 ST Array 10737962 - -
          Rat Genome 230 2.0 Array 1387813_at - -
          Rat Genome 230 2.0 Array 1387813_PM_at - -
          Affymetrix HT Rat Focus Array 1387813_PM_at - -
          Affymetrix HT Rat Focus Array 1387813_at - -
          Affymetrix Rat Clariom S Assay PSR1000013937.rn.1 TC1000001575.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000013938.rn.1 TC1000001575.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000013939.rn.1 TC1000001575.rn.1 RTA TC
          Rat Expression Set 230 1387813_PM_at - -
          Rat Expression Set 230 1387813_at - -
          Affymetrix Rat Gene 1.0 ST Array 10737962 - -
          Rat Genome U34 Set X03362_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21670625 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350020 - -
          RatRef-12 Expression BeadChip ILMN_69395 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline