Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566568_m1
          See other FH GE Assays ›
          Gene Symbol:
          Fh
          Gene Name:
          fumarate hydratase
          Gene Aliases:
          Fh1
          Chromosome Location:
          Chr.13: 93651486 - 93677371 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566568_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017005.2 NP_058701.2 6 - 7 907 80
          GenBank mRNA BC087598.1 - 6 - 7 907 7311498 80
          FQ218764.1 - 2 - 3 218 80
          FQ230216.1 - 2 - 3 252 80
          FQ232207.1 - 2 - 3 252 80
          J04473.1 - 6 - 7 918 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001997.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fh

          Panther Classification:

          Molecular Function -

          lyase

          Gene Ontology Categories:

          Function(s) Process(es)

          tricarboxylic acid cycle
          fumarate metabolic process
          malate metabolic process
          homeostasis of number of cells within a tissue
          fumarate hydratase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P642743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131649 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136076 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P642743 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P642743 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P642743 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P297250 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P545363 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P642743 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702080458 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701200421 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58866 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34950 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7363 - -
          Agilent Rat cDNA Microarray (G4105A) 702080458 - -
          Agilent Rat cDNA Microarray (G4105A) 701200421 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770147 - -
          Affymetrix Rat Gene 1.0 ST Array 10770145 - -
          Affymetrix Rat Gene 1.0 ST Array 10770148 - -
          Affymetrix Rat Gene 1.0 ST Array 10770152 - -
          Affymetrix Rat Gene 1.0 ST Array 10770144 - -
          Affymetrix Rat Gene 1.0 ST Array 10770146 - -
          Affymetrix Rat Gene 1.0 ST Array 10770153 - -
          Affymetrix Rat Gene 1.0 ST Array 10770151 - -
          Affymetrix Rat Gene 1.0 ST Array 10770149 - -
          Affymetrix Rat Gene 1.0 ST Array 10770150 - -
          Affymetrix Rat Gene 1.1 ST Array 10770143 - -
          Rat Genome 230 2.0 Array 1367670_at - -
          Rat Genome 230 2.0 Array 1367670_PM_at - -
          Affymetrix HT Rat Focus Array 1367670_at - -
          Affymetrix HT Rat Focus Array 1367670_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300014274.rn.1 TC1300001997.rn.1 RTA TC
          Rat Expression Set 230 1367670_PM_at - -
          Rat Expression Set 230 1367670_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770143 - -
          Rat Genome U34 Set rc_AI171734_s_at - -
          Rat Genome U34 Set J04473_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21129817 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364235 - -
          RatRef-12 Expression BeadChip ILMN_64247 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline