Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566881_m1
          See other PTGS1 GE Assays ›
          Gene Symbol:
          Ptgs1
          Gene Name:
          prostaglandin-endoperoxide synthase 1
          Gene Aliases:
          Cox-1, Cox-3, Cox1, Cox3, Pghs-1
          Chromosome Location:
          Chr.3: 15560685 - 15582339 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566881_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptgs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017043.4 NP_058739.4 9 - 10 1364 85
          XM_006234063.2 XP_006234125.1 9 - 10 1363 85
          GenBank mRNA BC081816.1 - 9 - 10 1364 7129759 85
          S67721.1 - 9 - 10 1357 85
          U03388.1 - 9 - 10 1357 85
          U18060.1 - 8 - 9 1305 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000322.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptgs1

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          prostaglandin metabolic process
          inflammatory response
          response to oxidative stress
          aging
          learning
          memory
          regulation of blood pressure
          response to organonitrogen compound
          negative regulation of norepinephrine secretion
          sensory perception of pain
          cyclooxygenase pathway
          negative regulation of epinephrine secretion
          maintenance of blood-brain barrier
          regulation of cell proliferation
          positive regulation of vasoconstriction
          positive regulation of smooth muscle contraction
          response to corticosterone
          oxidation-reduction process
          response to fatty acid
          peroxidase activity
          prostaglandin-endoperoxide synthase activity
          lipid binding
          heme binding
          metal ion binding
          dioxygenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P137448 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P375942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451639 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166559 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79024 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P137448 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P375942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P375942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P137448 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P375942 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P137448 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P137448 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701647331 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700594261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85256 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66499 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100671 - -
          Agilent Rat cDNA Microarray (G4105A) 700594261 - -
          Agilent Rat cDNA Microarray (G4105A) 701647331 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10835826 - -
          Affymetrix Rat Gene 1.0 ST Array 10835818 - -
          Affymetrix Rat Gene 1.0 ST Array 10835828 - -
          Affymetrix Rat Gene 1.0 ST Array 10835825 - -
          Affymetrix Rat Gene 1.0 ST Array 10835829 - -
          Affymetrix Rat Gene 1.0 ST Array 10835822 - -
          Affymetrix Rat Gene 1.0 ST Array 10835830 - -
          Affymetrix Rat Gene 1.0 ST Array 10835823 - -
          Affymetrix Rat Gene 1.0 ST Array 10835824 - -
          Affymetrix Rat Gene 1.0 ST Array 10835819 - -
          Affymetrix Rat Gene 1.0 ST Array 10835821 - -
          Affymetrix Rat Gene 1.0 ST Array 10835820 - -
          Affymetrix Rat Gene 1.0 ST Array 10835827 - -
          Affymetrix Rat Gene 1.1 ST Array 10835817 - -
          Rat Genome 230 2.0 Array 1381806_PM_at - -
          Rat Genome 230 2.0 Array 1368259_PM_at - -
          Rat Genome 230 2.0 Array 1381806_at - -
          Rat Genome 230 2.0 Array 1368259_at - -
          Affymetrix HT Rat Focus Array 1381806_at - -
          Affymetrix HT Rat Focus Array 1368259_at - -
          Affymetrix HT Rat Focus Array 1368259_PM_at - -
          Affymetrix HT Rat Focus Array 1381806_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300003565.rn.1 TC0300000322.rn.1 RTA TC
          Rat Expression Set 230 1368259_at - -
          Rat Expression Set 230 1368259_PM_at - -
          Rat Expression Set 230 1381806_at - -
          Rat Expression Set 230 1381806_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10835817 - -
          Rat Genome U34 Set U03388_s_at - -
          Rat Genome U34 Set U03388_s_at - -
          Rat Genome U34 Set U03388_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21300420 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0