Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567155_m1
          See other CHRNA5 GE Assays ›
          Gene Symbol:
          Chrna5
          Gene Name:
          cholinergic receptor, nicotinic, alpha 5 (neuronal)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 59561817 - 59590172 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567155_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chrna5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017078.2 NP_058774.2 1 - 2 169 145
          GenBank mRNA J05231.1 - 1 - 2 168 145

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the J05231.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chrna5

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          chemical synaptic transmission
          response to nicotine
          behavioral response to nicotine
          cation transmembrane transport
          acetylcholine-activated cation-selective channel activity
          acetylcholine receptor activity
          acetylcholine binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174662 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98445 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15448 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15448 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15448 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15448 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22012 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16193 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20814 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10910138 - -
          Affymetrix Rat Gene 1.0 ST Array 10910142 - -
          Affymetrix Rat Gene 1.0 ST Array 10910134 - -
          Affymetrix Rat Gene 1.0 ST Array 10910135 - -
          Affymetrix Rat Gene 1.0 ST Array 10910137 - -
          Affymetrix Rat Gene 1.0 ST Array 10917598 - -
          Affymetrix Rat Gene 1.0 ST Array 10910136 - -
          Affymetrix Rat Gene 1.0 ST Array 10910139 - -
          Affymetrix Rat Gene 1.0 ST Array 10910140 - -
          Affymetrix Rat Gene 1.0 ST Array 10910141 - -
          Affymetrix Rat Gene 1.1 ST Array 10910133 - -
          Rat Genome 230 2.0 Array 1369019_at - -
          Rat Genome 230 2.0 Array 1369019_PM_at - -
          Affymetrix HT Rat Focus Array 1369019_PM_at - -
          Affymetrix HT Rat Focus Array 1369019_at - -
          Affymetrix Rat Clariom S Assay PSR0800004998.rn.1 TC0800000745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800004999.rn.1 TC0800000745.rn.1 RTA TC
          Rat Expression Set 230 1369019_at - -
          Rat Expression Set 230 1369019_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10910133 - -
          Rat Genome U34 Set J05231_at - -
          Rat Genome U34 Set J05231_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22417456 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038816 - -
          RatRef-12 Expression BeadChip ILMN_53890 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline