Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567180_m1
          See other UMOD GE Assays ›
          Gene Symbol:
          Umod
          Gene Name:
          uromodulin
          Gene Aliases:
          THP
          Chromosome Location:
          Chr.1: 189186027 - 189199939 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567180_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Umod
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017082.1 NP_058778.1 9 - 10 1896 58
          XM_006230107.2 XP_006230169.1 10 - 11 2018 58
          GenBank mRNA BC081814.1 - 9 - 10 1902 7109450 58
          M63510.1 - 9 - 10 1896 58
          S75960.1 - 9 - 10 1887 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006507.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Umod

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          leukocyte cell-cell adhesion
          excretion
          response to organic substance
          chemical homeostasis
          metanephric ascending thin limb development
          metanephric distal convoluted tubule development
          metanephric thick ascending limb development
          neutrophil migration
          regulation of ion homeostasis
          calcium ion binding
          protein binding
          IgG binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872409 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6535 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69855 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022545 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022545 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022545 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11809 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701718156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34813 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24638 - -
          Agilent Rat cDNA Microarray (G4105A) 701718156 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725307 - -
          Affymetrix Rat Gene 1.0 ST Array 10725312 - -
          Affymetrix Rat Gene 1.0 ST Array 10725303 - -
          Affymetrix Rat Gene 1.0 ST Array 10725306 - -
          Affymetrix Rat Gene 1.0 ST Array 10725310 - -
          Affymetrix Rat Gene 1.0 ST Array 10725311 - -
          Affymetrix Rat Gene 1.0 ST Array 10725305 - -
          Affymetrix Rat Gene 1.0 ST Array 10725309 - -
          Affymetrix Rat Gene 1.0 ST Array 10725304 - -
          Affymetrix Rat Gene 1.0 ST Array 10725308 - -
          Affymetrix Rat Gene 1.1 ST Array 10725302 - -
          Rat Genome 230 2.0 Array 1368333_PM_at - -
          Rat Genome 230 2.0 Array 1368333_at - -
          Affymetrix HT Rat Focus Array 1368333_PM_at - -
          Affymetrix HT Rat Focus Array 1368333_at - -
          Affymetrix Rat Clariom S Assay PSR0100051086.rn.1 TC0100006507.rn.1 RTA TC
          Rat Expression Set 230 1368333_PM_at - -
          Rat Expression Set 230 1368333_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725302 - -
          Rat Genome U34 Set S75960_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20749968 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363631 - -
          RatRef-12 Expression BeadChip ILMN_53539 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline