Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567281_m1
          See other TYRO3 GE Assays ›
          Gene Symbol:
          Tyro3
          Gene Name:
          TYRO3 protein tyrosine kinase
          Gene Aliases:
          Brt
          Chromosome Location:
          Chr.3: 111597102 - 111616142 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567281_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tyro3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017092.1 NP_058788.1 7 - 8 955 72
          GenBank mRNA D37880.1 - 7 - 8 955 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001522.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tyro3

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          natural killer cell differentiation
          protein phosphorylation
          spermatogenesis
          peptidyl-tyrosine phosphorylation
          forebrain cell migration
          platelet activation
          secretion by cell
          negative regulation of toll-like receptor signaling pathway
          substrate adhesion-dependent cell spreading
          ovulation cycle
          apoptotic cell clearance
          protein kinase B signaling
          negative regulation of neuron apoptotic process
          negative regulation of innate immune response
          protein autophosphorylation
          negative regulation of inflammatory response
          negative regulation of lymphocyte activation
          vagina development
          neuron cellular homeostasis
          platelet aggregation
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          ATP binding
          phosphatidylinositol 3-kinase binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P065388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173290 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065388 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065388 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P065388 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P337455 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702080288 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50560 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74753 - -
          Agilent Rat cDNA Microarray (G4105A) 702080288 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10838901 - -
          Affymetrix Rat Gene 1.0 ST Array 10838886 - -
          Affymetrix Rat Gene 1.0 ST Array 10838894 - -
          Affymetrix Rat Gene 1.0 ST Array 10838900 - -
          Affymetrix Rat Gene 1.0 ST Array 10838885 - -
          Affymetrix Rat Gene 1.0 ST Array 10838896 - -
          Affymetrix Rat Gene 1.0 ST Array 10838897 - -
          Affymetrix Rat Gene 1.0 ST Array 10838889 - -
          Affymetrix Rat Gene 1.0 ST Array 10838892 - -
          Affymetrix Rat Gene 1.0 ST Array 10838883 - -
          Affymetrix Rat Gene 1.0 ST Array 10838888 - -
          Affymetrix Rat Gene 1.0 ST Array 10838899 - -
          Affymetrix Rat Gene 1.0 ST Array 10838898 - -
          Affymetrix Rat Gene 1.0 ST Array 10838891 - -
          Affymetrix Rat Gene 1.0 ST Array 10838890 - -
          Affymetrix Rat Gene 1.0 ST Array 10838895 - -
          Affymetrix Rat Gene 1.0 ST Array 10838887 - -
          Affymetrix Rat Gene 1.0 ST Array 10838884 - -
          Affymetrix Rat Gene 1.0 ST Array 10838893 - -
          Affymetrix Rat Gene 1.1 ST Array 10838881 - -
          Rat Genome 230 2.0 Array 1367953_PM_at - -
          Rat Genome 230 2.0 Array 1367953_at - -
          Affymetrix HT Rat Focus Array 1367953_at - -
          Affymetrix HT Rat Focus Array 1367953_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300010347.rn.1 TC0300001522.rn.1 RTA TC
          Rat Expression Set 230 1367953_at - -
          Rat Expression Set 230 1367953_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10838881 - -
          Rat Genome U34 Set D37880_at - -
          Rat Genome U34 Set rc_AA852055_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21478985 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358278 - -
          RatRef-12 Expression BeadChip ILMN_54221 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline