Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567471_m1
          See other AREG GE Assays ›
          Gene Symbol:
          Areg
          Gene Name:
          amphiregulin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 18521921 - 18531179 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567471_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Areg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017123.1 NP_058819.1 4 - 5 818 84
          GenBank mRNA X55183.1 - 4 - 5 818 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001499.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Areg

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          epidermal growth factor receptor signaling pathway
          G-protein coupled receptor signaling pathway
          central nervous system development
          positive regulation of cell proliferation
          glial cell proliferation
          response to organic cyclic compound
          neuron projection development
          response to estradiol
          positive regulation of phosphorylation
          response to hydrogen peroxide
          response to peptide hormone
          negative regulation of osteoblast differentiation
          positive regulation of DNA replication
          response to glucocorticoid
          response to cAMP
          dichotomous subdivision of terminal units involved in mammary gland duct morphogenesis
          mammary gland branching involved in thelarche
          mammary gland alveolus development
          epithelial cell proliferation involved in mammary gland duct elongation
          cytokine activity
          epidermal growth factor receptor binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P791677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732081 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162658 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P791677 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P791677 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P791677 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P791677 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27924 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775869 - -
          Affymetrix Rat Gene 1.0 ST Array 10775868 - -
          Affymetrix Rat Gene 1.0 ST Array 10775871 - -
          Affymetrix Rat Gene 1.0 ST Array 10775870 - -
          Affymetrix Rat Gene 1.0 ST Array 10775872 - -
          Affymetrix Rat Gene 1.0 ST Array 10775867 - -
          Affymetrix Rat Gene 1.1 ST Array 10775866 - -
          Rat Genome 230 2.0 Array 1369871_at - -
          Rat Genome 230 2.0 Array 1369871_PM_at - -
          Affymetrix HT Rat Focus Array 1369871_at - -
          Affymetrix HT Rat Focus Array 1369871_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400011010.rn.1 TC1400001499.rn.1 RTA TC
          Rat Expression Set 230 1369871_PM_at - -
          Rat Expression Set 230 1369871_at - -
          Affymetrix Rat Gene 1.0 ST Array 10775866 - -
          Rat Genome U34 Set X55183_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21390466 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363074 - -
          RatRef-12 Expression BeadChip ILMN_65820 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline