Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567505_m1
          See other NEU2 GE Assays ›
          Gene Symbol:
          Neu2
          Gene Name:
          sialidase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 94702095 - 94724903 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567505_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Neu2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017130.2 NP_058826.2 2 - 3 396 59
          XM_006245361.2 XP_006245423.1 2 - 3 336 59
          XM_006245362.2 XP_006245424.1 2 - 3 1000 59
          XM_006245363.2 XP_006245425.1 2 - 3 347 59
          XM_006245364.1 XP_006245426.1 2 - 3 302 59
          XM_006245365.2 XP_006245427.1 2 - 3 343 59
          XM_008767209.1 XP_008765431.1 3 - 4 528 59
          XM_008767210.1 XP_008765432.1 3 - 4 474 59
          XM_008767211.1 XP_008765433.1 2 - 3 482 59
          GenBank mRNA BC091154.1 - 2 - 3 325 7323285 59
          D16300.1 - 2 - 3 396 59
          FQ213221.1 - 2 - 3 359 59
          FQ216178.1 - 2 - 3 271 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001080.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Neu2

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          ganglioside catabolic process
          oligosaccharide catabolic process
          positive regulation of myotube differentiation
          response to ethanol
          positive regulation of myoblast differentiation
          exo-alpha-sialidase activity
          exo-alpha-(2->3)-sialidase activity
          exo-alpha-(2->6)-sialidase activity
          exo-alpha-(2->8)-sialidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P467591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594246 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P467591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P467591 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P467591 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P467591 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700917592 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75377 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93517 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89714 - -
          Agilent Rat cDNA Microarray (G4105A) 700917592 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925057 - -
          Affymetrix Rat Gene 1.0 ST Array 10925059 - -
          Affymetrix Rat Gene 1.0 ST Array 10925058 - -
          Affymetrix Rat Gene 1.1 ST Array 10925055 - -
          Rat Genome 230 2.0 Array 1368127_at - -
          Rat Genome 230 2.0 Array 1368127_PM_at - -
          Affymetrix HT Rat Focus Array 1368127_at - -
          Affymetrix HT Rat Focus Array 1368127_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900009197.rn.1 TC0900001080.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900009209.rn.1 TC0900001080.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900009208.rn.1 TC0900001080.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900009198.rn.1 TC0900001080.rn.1 RTA TC
          Rat Expression Set 230 1368127_PM_at - -
          Rat Expression Set 230 1368127_at - -
          Affymetrix Rat Gene 1.0 ST Array 10925055 - -
          Rat Genome U34 Set rc_AI171959_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21591663 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375230 - -
          RatRef-12 Expression BeadChip ILMN_57812 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline