Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567682_m1
          See other HAL GE Assays ›
          Gene Symbol:
          Hal
          Gene Name:
          histidine ammonia lyase
          Gene Aliases:
          histidase
          Chromosome Location:
          Chr.7: 34326087 - 34356413 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567682_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hal
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017159.1 NP_058855.1 17 - 18 1662 63
          GenBank mRNA BC089809.1 - 17 - 18 1691 7325816 63
          M58308.1 - 17 - 18 1662 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000514.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hal

          Panther Classification:

          Molecular Function -

          lyase

          Gene Ontology Categories:

          Function(s) Process(es)

          histidine metabolic process
          histidine catabolic process
          biosynthetic process
          histidine catabolic process to glutamate and formamide
          histidine catabolic process to glutamate and formate
          histidine ammonia-lyase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1038341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351988 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25050 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171836 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34884 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1038341 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1038341 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1038341 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P245474 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1038341 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701580003 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50731 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47239 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83455 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38376 - -
          Agilent Rat cDNA Microarray (G4105A) 701580003 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10894911 - -
          Affymetrix Rat Gene 1.0 ST Array 10894908 - -
          Affymetrix Rat Gene 1.0 ST Array 10894902 - -
          Affymetrix Rat Gene 1.0 ST Array 10894917 - -
          Affymetrix Rat Gene 1.0 ST Array 10894907 - -
          Affymetrix Rat Gene 1.0 ST Array 10894916 - -
          Affymetrix Rat Gene 1.0 ST Array 10894912 - -
          Affymetrix Rat Gene 1.0 ST Array 10894918 - -
          Affymetrix Rat Gene 1.0 ST Array 10894905 - -
          Affymetrix Rat Gene 1.0 ST Array 10894915 - -
          Affymetrix Rat Gene 1.0 ST Array 10894909 - -
          Affymetrix Rat Gene 1.0 ST Array 10894913 - -
          Affymetrix Rat Gene 1.0 ST Array 10894901 - -
          Affymetrix Rat Gene 1.0 ST Array 10894910 - -
          Affymetrix Rat Gene 1.0 ST Array 10894903 - -
          Affymetrix Rat Gene 1.0 ST Array 10894914 - -
          Affymetrix Rat Gene 1.0 ST Array 10894899 - -
          Affymetrix Rat Gene 1.0 ST Array 10894898 - -
          Affymetrix Rat Gene 1.0 ST Array 10894900 - -
          Affymetrix Rat Gene 1.0 ST Array 10894906 - -
          Affymetrix Rat Gene 1.0 ST Array 10894904 - -
          Affymetrix Rat Gene 1.1 ST Array 10894897 - -
          Rat Genome 230 2.0 Array 1387307_PM_at - -
          Rat Genome 230 2.0 Array 1387307_at - -
          Affymetrix HT Rat Focus Array 1387307_PM_at - -
          Affymetrix HT Rat Focus Array 1387307_at - -
          Affymetrix Rat Clariom S Assay PSR0700004278.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004280.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004291.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004282.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004277.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004281.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004284.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004287.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004289.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004292.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004295.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004275.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004279.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004283.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004285.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004293.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004276.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004286.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004288.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004290.rn.1 TC0700000514.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004294.rn.1 TC0700000514.rn.1 RTA TC
          Rat Expression Set 230 1387307_at - -
          Rat Expression Set 230 1387307_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10894897 - -
          Rat Genome U34 Set AB002393_g_at - -
          Rat Genome U34 Set M58308_at - -
          Rat Genome U34 Set AB002393_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21787755 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364209 - -
          RatRef-12 Expression BeadChip ILMN_54598 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline