Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567714_m1
          See other SLC6A18 GE Assays ›
          Gene Symbol:
          Slc6a18
          Gene Name:
          solute carrier family 6 (neutral amino acid transporter), member 18
          Gene Aliases:
          Rosit, Xtrp2
          Chromosome Location:
          Chr.1: 32221680 - 32235599 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567714_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc6a18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017163.1 NP_058859.1 7 - 8 1002 79
          XM_006227772.2 XP_006227834.1 7 - 8 1090 79
          XM_006227774.2 XP_006227836.1 7 - 8 1090 79
          XM_006227775.2 XP_006227837.1 7 - 8 1090 79
          XM_008758682.1 XP_008756904.1 6 - 7 979 79
          GenBank mRNA U12973.1 - 7 - 8 1002 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000340.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc6a18

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transmembrane transport
          neurotransmitter transport
          amino acid transport
          response to osmotic stress
          hyperosmotic response
          transmembrane transport
          neurotransmitter:sodium symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11843 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P128852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307009 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66429 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38664 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11843 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11843 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11843 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P128852 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P132711 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24085 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702569 - -
          Affymetrix Rat Gene 1.0 ST Array 10702571 - -
          Affymetrix Rat Gene 1.0 ST Array 10702572 - -
          Affymetrix Rat Gene 1.0 ST Array 10702564 - -
          Affymetrix Rat Gene 1.0 ST Array 10702567 - -
          Affymetrix Rat Gene 1.0 ST Array 10702568 - -
          Affymetrix Rat Gene 1.0 ST Array 10702566 - -
          Affymetrix Rat Gene 1.0 ST Array 10702570 - -
          Affymetrix Rat Gene 1.0 ST Array 10702577 - -
          Affymetrix Rat Gene 1.0 ST Array 10702576 - -
          Affymetrix Rat Gene 1.0 ST Array 10702578 - -
          Affymetrix Rat Gene 1.0 ST Array 10702574 - -
          Affymetrix Rat Gene 1.0 ST Array 10702565 - -
          Affymetrix Rat Gene 1.0 ST Array 10702563 - -
          Affymetrix Rat Gene 1.0 ST Array 10702575 - -
          Affymetrix Rat Gene 1.1 ST Array 10702562 - -
          Rat Genome 230 2.0 Array 1368575_PM_at - -
          Rat Genome 230 2.0 Array 1368575_at - -
          Affymetrix HT Rat Focus Array 1368575_at - -
          Affymetrix HT Rat Focus Array 1368575_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100002029.rn.1 TC0100000340.rn.1 RTA TC
          Rat Expression Set 230 1368575_PM_at - -
          Rat Expression Set 230 1368575_at - -
          Affymetrix Rat Gene 1.0 ST Array 10702562 - -
          Rat Genome U34 Set U12973_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22152223 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353373 - -
          RatRef-12 Expression BeadChip ILMN_50239 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline