Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567818_m1
          See other BMP2 GE Assays ›
          Gene Symbol:
          Bmp2
          Gene Name:
          bone morphogenetic protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 126335963 - 126345881 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567818_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bmp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017178.1 NP_058874.1 2 - 3 408 126
          XM_008762246.1 XP_008760468.1 1 - 2 1677 126
          GenBank mRNA L20678.1 - 2 - 3 408 126
          Z25868.1 - 2 - 3 408 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001785.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bmp2

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPK activity
          ossification
          osteoblast differentiation
          branching involved in ureteric bud morphogenesis
          response to hypoxia
          in utero embryonic development
          epithelial to mesenchymal transition
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          chondrocyte differentiation
          BMP signaling pathway involved in heart induction
          atrioventricular valve morphogenesis
          endocardial cushion morphogenesis
          negative regulation of Wnt signaling pathway involved in heart development
          proteoglycan metabolic process
          regulation of transcription, DNA-templated
          protein phosphorylation
          inflammatory response
          Notch signaling pathway
          heart development
          negative regulation of cell proliferation
          response to mechanical stimulus
          organ morphogenesis
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of steroid biosynthetic process
          positive regulation of phosphatase activity
          telencephalon development
          telencephalon regionalization
          positive regulation of Wnt signaling pathway
          bone mineralization
          positive regulation of cell migration
          positive regulation of bone mineralization
          BMP signaling pathway
          protein destabilization
          negative regulation of aldosterone biosynthetic process
          response to retinoic acid
          positive regulation of osteoblast proliferation
          cardiocyte differentiation
          embryonic heart tube anterior/posterior pattern specification
          bone mineralization involved in bone maturation
          growth
          odontogenesis of dentin-containing tooth
          positive regulation of odontogenesis
          regulation of odontogenesis of dentin-containing tooth
          ovulation cycle
          regulation of apoptotic process
          positive regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of MAPK cascade
          negative regulation of insulin-like growth factor receptor signaling pathway
          cell fate commitment
          positive regulation of cell differentiation
          positive regulation of fat cell differentiation
          positive regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of ossification
          negative regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of astrocyte differentiation
          mesenchymal cell differentiation
          inner ear development
          positive regulation of neurogenesis
          negative regulation of calcium-independent cell-cell adhesion
          cartilage development
          cardiac muscle cell differentiation
          cardiac muscle tissue morphogenesis
          oxidation-reduction process
          pericardium development
          corticotropin hormone secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          cardiac epithelial to mesenchymal transition
          pathway-restricted SMAD protein phosphorylation
          SMAD protein signal transduction
          mesenchyme development
          positive regulation of Wnt signaling pathway by BMP signaling pathway
          positive regulation of cartilage development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to mechanical stimulus
          cellular response to growth factor stimulus
          cellular response to organic cyclic compound
          cellular response to BMP stimulus
          mesenchymal cell proliferation involved in ureteric bud development
          negative regulation of canonical Wnt signaling pathway
          positive regulation of p38MAPK cascade
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          negative regulation of cortisol biosynthetic process
          negative regulation of cardiac muscle cell differentiation
          retinol dehydrogenase activity
          receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          growth factor activity
          phosphatase activator activity
          protein domain specific binding
          protein homodimerization activity
          SMAD binding
          protein heterodimerization activity
          BMP receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1013851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21638 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54257 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1013851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1013851 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1013851 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11848 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1013851 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700637909 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43083 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43029 - -
          Agilent Rat cDNA Microarray (G4105A) 700637909 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840141 - -
          Affymetrix Rat Gene 1.0 ST Array 10840139 - -
          Affymetrix Rat Gene 1.0 ST Array 10840140 - -
          Affymetrix Rat Gene 1.1 ST Array 10840138 - -
          Rat Genome 230 2.0 Array 1368945_at - -
          Rat Genome 230 2.0 Array 1398270_PM_at - -
          Rat Genome 230 2.0 Array 1398270_at - -
          Rat Genome 230 2.0 Array 1368945_PM_at - -
          Affymetrix HT Rat Focus Array 1398270_PM_at - -
          Affymetrix HT Rat Focus Array 1368945_PM_at - -
          Affymetrix HT Rat Focus Array 1368945_at - -
          Affymetrix HT Rat Focus Array 1398270_at - -
          Affymetrix Rat Clariom S Assay PSR0300012896.rn.1 TC0300001785.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300012897.rn.1 TC0300001785.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300012901.rn.1 TC0300001785.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300012900.rn.1 TC0300001785.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300012898.rn.1 TC0300001785.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300012899.rn.1 TC0300001785.rn.1 RTA TC
          Rat Expression Set 230 1368945_PM_at - -
          Rat Expression Set 230 1398270_at - -
          Rat Expression Set 230 1368945_at - -
          Rat Expression Set 230 1398270_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840138 - -
          Rat Genome U34 Set rc_AA997410_s_at - -
          Rat Genome U34 Set L20678_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21743176 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356614 - -
          RatRef-12 Expression BeadChip ILMN_63615 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline