Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567985_m1
          See other LBP GE Assays ›
          Gene Symbol:
          Lbp
          Gene Name:
          lipopolysaccharide binding protein
          Gene Aliases:
          Bpifd2
          Chromosome Location:
          Chr.3: 154786232 - 154812910 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567985_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lbp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017208.2 NP_058904.2 1 - 2 147 70
          GenBank mRNA BC101906.1 - 1 - 2 147 7933709 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002191.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lbp

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          leukocyte chemotaxis involved in inflammatory response
          macrophage activation involved in immune response
          lipid transport
          acute-phase response
          lipopolysaccharide transport
          lipopolysaccharide-mediated signaling pathway
          detection of molecule of bacterial origin
          response to lipopolysaccharide
          negative regulation of tumor necrosis factor production
          positive regulation of chemokine production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of toll-like receptor 4 signaling pathway
          positive regulation of tumor necrosis factor biosynthetic process
          positive regulation of macrophage activation
          negative regulation of growth of symbiont in host
          innate immune response
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          positive regulation of phagocytosis, engulfment
          positive regulation of respiratory burst involved in inflammatory response
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          positive regulation of neutrophil chemotaxis
          regulation of membrane permeability
          lipopolysaccharide binding
          receptor binding
          lipoteichoic acid binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P468468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91127 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99134 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P468468 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P468468 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P468468 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P468468 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37587 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841696 - -
          Affymetrix Rat Gene 1.0 ST Array 10841698 - -
          Affymetrix Rat Gene 1.0 ST Array 10841697 - -
          Affymetrix Rat Gene 1.0 ST Array 10841700 - -
          Affymetrix Rat Gene 1.0 ST Array 10841707 - -
          Affymetrix Rat Gene 1.0 ST Array 10841699 - -
          Affymetrix Rat Gene 1.0 ST Array 10841704 - -
          Affymetrix Rat Gene 1.0 ST Array 10841703 - -
          Affymetrix Rat Gene 1.0 ST Array 10841705 - -
          Affymetrix Rat Gene 1.0 ST Array 10841706 - -
          Affymetrix Rat Gene 1.0 ST Array 10841708 - -
          Affymetrix Rat Gene 1.0 ST Array 10841701 - -
          Affymetrix Rat Gene 1.0 ST Array 10841702 - -
          Affymetrix Rat Gene 1.0 ST Array 10841694 - -
          Affymetrix Rat Gene 1.0 ST Array 10841695 - -
          Affymetrix Rat Gene 1.1 ST Array 10841693 - -
          Rat Genome 230 2.0 Array 1387868_PM_at - -
          Rat Genome 230 2.0 Array 1387868_at - -
          Affymetrix HT Rat Focus Array 1387868_PM_at - -
          Affymetrix HT Rat Focus Array 1387868_at - -
          Affymetrix Rat Clariom S Assay PSR0300016219.rn.1 TC0300002191.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300016221.rn.1 TC0300002191.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300016220.rn.1 TC0300002191.rn.1 RTA TC
          Rat Expression Set 230 1387868_PM_at - -
          Rat Expression Set 230 1387868_at - -
          Affymetrix Rat Gene 1.0 ST Array 10841693 - -
          Rat Genome U34 Set L32132_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376661 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline