Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568021_m1
          See other GLG1 GE Assays ›
          Gene Symbol:
          Glg1
          Gene Name:
          golgi glycoprotein 1
          Gene Aliases:
          MG-160, Selel
          Chromosome Location:
          Chr.19: 42983879 - 43088074 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568021_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Glg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017211.2 NP_058907.1 10 - 11 1663 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000596.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Glg1

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          negative regulation of protein processing
          negative regulation of transforming growth factor beta receptor signaling pathway
          regulation of chondrocyte differentiation
          bone morphogenesis
          fibroblast growth factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P383965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171722 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118782 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101265 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146020 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171735 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133526 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P383965 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P383965 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P383965 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P383965 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40149 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98636 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75577 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103412 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29808 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81175 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52627 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811079 - -
          Affymetrix Rat Gene 1.0 ST Array 10811077 - -
          Affymetrix Rat Gene 1.0 ST Array 10811098 - -
          Affymetrix Rat Gene 1.0 ST Array 10811088 - -
          Affymetrix Rat Gene 1.0 ST Array 10811083 - -
          Affymetrix Rat Gene 1.0 ST Array 10811092 - -
          Affymetrix Rat Gene 1.0 ST Array 10811095 - -
          Affymetrix Rat Gene 1.0 ST Array 10811100 - -
          Affymetrix Rat Gene 1.0 ST Array 10811089 - -
          Affymetrix Rat Gene 1.0 ST Array 10811099 - -
          Affymetrix Rat Gene 1.0 ST Array 10811101 - -
          Affymetrix Rat Gene 1.0 ST Array 10811087 - -
          Affymetrix Rat Gene 1.0 ST Array 10811097 - -
          Affymetrix Rat Gene 1.0 ST Array 10811084 - -
          Affymetrix Rat Gene 1.0 ST Array 10811082 - -
          Affymetrix Rat Gene 1.0 ST Array 10811090 - -
          Affymetrix Rat Gene 1.0 ST Array 10811078 - -
          Affymetrix Rat Gene 1.0 ST Array 10811085 - -
          Affymetrix Rat Gene 1.0 ST Array 10811093 - -
          Affymetrix Rat Gene 1.0 ST Array 10811094 - -
          Affymetrix Rat Gene 1.0 ST Array 10811096 - -
          Affymetrix Rat Gene 1.0 ST Array 10811086 - -
          Affymetrix Rat Gene 1.0 ST Array 10811080 - -
          Affymetrix Rat Gene 1.0 ST Array 10811091 - -
          Affymetrix Rat Gene 1.1 ST Array 10811076 - -
          Rat Genome 230 2.0 Array 1387367_PM_at - -
          Rat Genome 230 2.0 Array 1387367_at - -
          Affymetrix HT Rat Focus Array 1387367_at - -
          Affymetrix HT Rat Focus Array 1387367_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900005069.rn.1 TC1900000596.rn.1 RTA TC
          Rat Expression Set 230 1387367_PM_at - -
          Rat Expression Set 230 1387367_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811076 - -
          Rat Genome U34 Set U08136_at - -
          Rat Genome U34 Set rc_AI176461_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21189934 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368524 - -
          RatRef-12 Expression BeadChip ILMN_68750 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline