Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568130_m1
          See other SLC20A2 GE Assays ›
          Gene Symbol:
          Slc20a2
          Gene Name:
          solute carrier family 20 (phosphate transporter), member 2
          Gene Aliases:
          Ab1-188, Glvr2, Pit-2, RAM-1, Ram1
          Chromosome Location:
          Chr.16: 74318287 - 74408030 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568130_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc20a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017223.2 NP_058919.1 2 - 3 703 96
          XM_008771351.1 XP_008769573.1 2 - 3 1710 96
          XM_008771352.1 XP_008769574.1 2 - 3 1350 96
          GenBank mRNA BC070908.1 - 2 - 3 703 7103640 96
          L19931.1 - 1 - 2 559 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001916.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc20a2

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          phosphate ion transport
          viral process
          transmembrane transport
          inorganic phosphate transmembrane transporter activity
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P078666 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P078664 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124726 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42063 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88043 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10047 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078666 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078664 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10047 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078666 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078664 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P078664 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P078666 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10047 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P449774 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65750 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56881 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51393 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792444 - -
          Affymetrix Rat Gene 1.0 ST Array 10792445 - -
          Affymetrix Rat Gene 1.0 ST Array 10792443 - -
          Affymetrix Rat Gene 1.0 ST Array 10792446 - -
          Affymetrix Rat Gene 1.0 ST Array 10792447 - -
          Affymetrix Rat Gene 1.1 ST Array 10792441 - -
          Rat Genome 230 2.0 Array 1372747_at - -
          Rat Genome 230 2.0 Array 1369234_PM_at - -
          Rat Genome 230 2.0 Array 1369234_at - -
          Rat Genome 230 2.0 Array 1372747_PM_at - -
          Affymetrix HT Rat Focus Array 1372747_PM_at - -
          Affymetrix HT Rat Focus Array 1369234_PM_at - -
          Affymetrix HT Rat Focus Array 1369234_at - -
          Affymetrix HT Rat Focus Array 1372747_at - -
          Affymetrix Rat Clariom S Assay PSR1600014170.rn.1 TC1600001916.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600014168.rn.1 TC1600001916.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600014169.rn.1 TC1600001916.rn.1 RTA TC
          Rat Expression Set 230 1372747_PM_at - -
          Rat Expression Set 230 1369234_PM_at - -
          Rat Expression Set 230 1369234_at - -
          Rat Expression Set 230 1372747_at - -
          Affymetrix Rat Gene 1.0 ST Array 10792441 - -
          Rat Genome U34 Set L19931_at - -
          Rat Genome U34 Set rc_AA923955_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21161567 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370678 - -
          RatRef-12 Expression BeadChip ILMN_56458 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline