Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568222_m1
          See other PITPNA GE Assays ›
          Gene Symbol:
          Pitpna
          Gene Name:
          phosphatidylinositol transfer protein, alpha
          Gene Aliases:
          Pitpn
          Chromosome Location:
          Chr.10: 63731767 - 63772049 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568222_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pitpna
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017231.1 NP_058927.1 7 - 8 656 87
          GenBank mRNA BC070945.1 - 7 - 8 684 7103390 87
          FQ235021.1 - 1 - 2 57 87
          M25758.1 - 7 - 8 656 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001112.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pitpna

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          phospholipid transport
          fatty-acyl-CoA binding
          phospholipid binding
          phosphatidylinositol transporter activity
          phosphatidylinositol binding
          stearic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P484902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22452 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117988 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81097 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P484902 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P484902 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P484902 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P484902 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46401 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75012 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41967 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735981 - -
          Affymetrix Rat Gene 1.0 ST Array 10735975 - -
          Affymetrix Rat Gene 1.0 ST Array 10735982 - -
          Affymetrix Rat Gene 1.0 ST Array 10735979 - -
          Affymetrix Rat Gene 1.0 ST Array 10735985 - -
          Affymetrix Rat Gene 1.0 ST Array 10735977 - -
          Affymetrix Rat Gene 1.0 ST Array 10735976 - -
          Affymetrix Rat Gene 1.0 ST Array 10735980 - -
          Affymetrix Rat Gene 1.0 ST Array 10735984 - -
          Affymetrix Rat Gene 1.0 ST Array 10735983 - -
          Affymetrix Rat Gene 1.0 ST Array 10735978 - -
          Affymetrix Rat Gene 1.1 ST Array 10735974 - -
          Rat Genome 230 2.0 Array 1398806_PM_at - -
          Rat Genome 230 2.0 Array 1398806_at - -
          Affymetrix HT Rat Focus Array 1398806_PM_at - -
          Affymetrix HT Rat Focus Array 1398806_at - -
          Affymetrix Rat Clariom S Assay PSR1000010087.rn.1 TC1000001112.rn.1 RTA TC
          Rat Expression Set 230 1398806_at - -
          Rat Expression Set 230 1398806_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10735974 - -
          Rat Genome U34 Set M25758_at - -
          Rat Genome U34 Set rc_AA956671_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21818973 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352480 - -
          RatRef-12 Expression BeadChip ILMN_69014 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline