Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568393_m1
          See other SCN10A GE Assays ›
          Gene Symbol:
          Scn10a
          Gene Name:
          sodium channel, voltage-gated, type X, alpha subunit
          Gene Aliases:
          Na(V)1.8, PN3
          Chromosome Location:
          Chr.8: 128298593 - 128416897 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568393_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scn10a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017247.1 NP_058943.1 14 - 15 2306 69
          XM_008766673.1 XP_008764895.1 14 - 15 2354 69
          GenBank mRNA X92184.1 - 14 - 15 2306 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003551.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scn10a

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          sodium ion transport
          neuronal action potential
          sensory perception of pain
          regulation of ion transmembrane transport
          sodium ion transmembrane transport
          odontogenesis of dentin-containing tooth
          regulation of cardiac muscle contraction
          regulation of atrial cardiac muscle cell membrane depolarization
          cardiac conduction
          membrane depolarization during action potential
          AV node cell to bundle of His cell communication
          bundle of His cell to Purkinje myocyte communication
          voltage-gated sodium channel activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P069374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375159 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71076 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65860 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75896 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84150 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150101 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148388 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99673 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36626 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167133 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94152 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150780 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37578 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069374 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069374 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P069374 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P278549 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5772 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3216 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23952 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21821 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23614 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86867 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 474 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65265 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10920916 - -
          Affymetrix Rat Gene 1.0 ST Array 10920907 - -
          Affymetrix Rat Gene 1.0 ST Array 10920923 - -
          Affymetrix Rat Gene 1.0 ST Array 10920928 - -
          Affymetrix Rat Gene 1.0 ST Array 10920922 - -
          Affymetrix Rat Gene 1.0 ST Array 10920925 - -
          Affymetrix Rat Gene 1.0 ST Array 10920914 - -
          Affymetrix Rat Gene 1.0 ST Array 10920920 - -
          Affymetrix Rat Gene 1.0 ST Array 10920918 - -
          Affymetrix Rat Gene 1.0 ST Array 10920927 - -
          Affymetrix Rat Gene 1.0 ST Array 10920903 - -
          Affymetrix Rat Gene 1.0 ST Array 10920905 - -
          Affymetrix Rat Gene 1.0 ST Array 10920902 - -
          Affymetrix Rat Gene 1.0 ST Array 10920926 - -
          Affymetrix Rat Gene 1.0 ST Array 10920908 - -
          Affymetrix Rat Gene 1.0 ST Array 10920921 - -
          Affymetrix Rat Gene 1.0 ST Array 10920912 - -
          Affymetrix Rat Gene 1.0 ST Array 10920904 - -
          Affymetrix Rat Gene 1.0 ST Array 10920913 - -
          Affymetrix Rat Gene 1.0 ST Array 10920917 - -
          Affymetrix Rat Gene 1.0 ST Array 10920911 - -
          Affymetrix Rat Gene 1.0 ST Array 10920919 - -
          Affymetrix Rat Gene 1.0 ST Array 10920906 - -
          Affymetrix Rat Gene 1.0 ST Array 10920915 - -
          Affymetrix Rat Gene 1.0 ST Array 10920910 - -
          Affymetrix Rat Gene 1.0 ST Array 10920924 - -
          Affymetrix Rat Gene 1.0 ST Array 10920909 - -
          Affymetrix Rat Gene 1.1 ST Array 10920900 - -
          Rat Genome 230 2.0 Array 1368351_at - -
          Rat Genome 230 2.0 Array 1368351_PM_at - -
          Affymetrix HT Rat Focus Array 1368351_at - -
          Affymetrix HT Rat Focus Array 1368351_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800028920.rn.1 TC0800003551.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800028922.rn.1 TC0800003551.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800028921.rn.1 TC0800003551.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800028918.rn.1 TC0800003551.rn.1 RTA TC
          Rat Expression Set 230 1368351_PM_at - -
          Rat Expression Set 230 1368351_at - -
          Affymetrix Rat Gene 1.0 ST Array 10920900 - -
          Rat Genome U34 Set X92184_at - -
          Rat Genome U34 Set X92184_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21865798 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362724 - -
          RatRef-12 Expression BeadChip ILMN_48343 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-xpdfw:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline