Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568482_m1
          See other TGFBR3 GE Assays ›
          Gene Symbol:
          Tgfbr3
          Gene Name:
          transforming growth factor, beta receptor III
          Gene Aliases:
          betaglycan
          Chromosome Location:
          Chr.14: 3506416 - 3680508 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568482_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfbr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017256.1 NP_058952.1 15 - 16 2669 89
          GenBank mRNA M77809.1 - 15 - 16 2669 89
          M80784.1 - 15 - 16 2654 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002457.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgfbr3

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          response to hypoxia
          in utero embryonic development
          blastocyst development
          epithelial to mesenchymal transition
          liver development
          protein complex assembly
          protein phosphorylation
          immune response
          signal transduction
          transforming growth factor beta receptor signaling pathway
          transforming growth factor beta receptor complex assembly
          negative regulation of epithelial cell migration
          negative regulation of epithelial to mesenchymal transition
          cell growth
          BMP signaling pathway
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          organ regeneration
          response to follicle-stimulating hormone
          response to prostaglandin E
          response to luteinizing hormone
          intracellular signal transduction
          regulation of protein binding
          negative regulation of epithelial cell proliferation
          positive regulation of NF-kappaB transcription factor activity
          negative regulation of cellular component movement
          ventricular cardiac muscle tissue morphogenesis
          palate development
          cardiac muscle cell proliferation
          definitive hemopoiesis
          cardiac epithelial to mesenchymal transition
          definitive erythrocyte differentiation
          heart trabecula formation
          pathway-restricted SMAD protein phosphorylation
          transforming growth factor beta-activated receptor activity
          type II transforming growth factor beta receptor binding
          transforming growth factor beta receptor binding
          protein binding
          glycosaminoglycan binding
          heparin binding
          coreceptor activity
          fibroblast growth factor binding
          PDZ domain binding
          SMAD binding
          activin binding
          transforming growth factor beta binding
          transforming growth factor beta receptor activity, type III

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P046313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569031 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115050 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54683 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8836 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106553 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48659 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110038 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113849 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178879 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116408 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6648 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89034 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91090 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9076 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046313 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046313 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P046313 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11883 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701827944 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700883378 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2819 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24477 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101631 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50923 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74341 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84765 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72889 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25641 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20997 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78152 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66852 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2492 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43543 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61551 - -
          Agilent Rat cDNA Microarray (G4105A) 701827944 - -
          Agilent Rat cDNA Microarray (G4105A) 700883378 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771088 - -
          Affymetrix Rat Gene 1.0 ST Array 10771075 - -
          Affymetrix Rat Gene 1.0 ST Array 10771072 - -
          Affymetrix Rat Gene 1.0 ST Array 10771084 - -
          Affymetrix Rat Gene 1.0 ST Array 10771073 - -
          Affymetrix Rat Gene 1.0 ST Array 10771087 - -
          Affymetrix Rat Gene 1.0 ST Array 10771081 - -
          Affymetrix Rat Gene 1.0 ST Array 10771085 - -
          Affymetrix Rat Gene 1.0 ST Array 10771080 - -
          Affymetrix Rat Gene 1.0 ST Array 10771082 - -
          Affymetrix Rat Gene 1.0 ST Array 10771074 - -
          Affymetrix Rat Gene 1.0 ST Array 10771077 - -
          Affymetrix Rat Gene 1.0 ST Array 10771076 - -
          Affymetrix Rat Gene 1.0 ST Array 10771078 - -
          Affymetrix Rat Gene 1.0 ST Array 10771086 - -
          Affymetrix Rat Gene 1.0 ST Array 10771079 - -
          Affymetrix Rat Gene 1.0 ST Array 10771071 - -
          Affymetrix Rat Gene 1.0 ST Array 10771083 - -
          Affymetrix Rat Gene 1.1 ST Array 10771070 - -
          Rat Genome 230 2.0 Array 1369219_PM_at - -
          Rat Genome 230 2.0 Array 1387484_PM_at - -
          Rat Genome 230 2.0 Array 1369219_at - -
          Rat Genome 230 2.0 Array 1387484_at - -
          Affymetrix HT Rat Focus Array 1387484_PM_at - -
          Affymetrix HT Rat Focus Array 1369219_PM_at - -
          Affymetrix HT Rat Focus Array 1387484_at - -
          Affymetrix HT Rat Focus Array 1369219_at - -
          Affymetrix Rat Clariom S Assay PSR1400018202.rn.1 TC1400002457.rn.1 RTA TC
          Rat Expression Set 230 1369219_at - -
          Rat Expression Set 230 1369219_PM_at - -
          Rat Expression Set 230 1387484_PM_at - -
          Rat Expression Set 230 1387484_at - -
          Affymetrix Rat Gene 1.0 ST Array 10771070 - -
          Rat Genome U34 Set M80784_s_at - -
          Rat Genome U34 Set M77809_at - -
          Rat Genome U34 Set M80784_s_at - -
          Rat Genome U34 Set M77809_at - -
          Rat Genome U34 Set M77809_at - -
          Rat Genome U34 Set M80784_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21536839 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354031 - -
          RatRef-12 Expression BeadChip ILMN_57170 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline