Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568506_m1
          See other ALOX5AP GE Assays ›
          Gene Symbol:
          Alox5ap
          Gene Name:
          arachidonate 5-lipoxygenase activating protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 6854930 - 6879112 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568506_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Alox5ap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017260.2 NP_058956.1 2 - 3 223 74
          GenBank mRNA BC086593.1 - 2 - 3 223 7310271 74
          X52196.1 - 2 - 3 205 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001002.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Alox5ap

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          leukotriene production involved in inflammatory response
          positive regulation of acute inflammatory response
          leukotriene biosynthetic process
          lipoxygenase pathway
          positive regulation of catalytic activity
          oxidation-reduction process
          protein homotrimerization
          cellular response to calcium ion
          arachidonate 5-lipoxygenase activity
          glutathione transferase activity
          leukotriene-C4 synthase activity
          glutathione peroxidase activity
          enzyme activator activity
          enzyme binding
          protein homodimerization activity
          protein heterodimerization activity
          protein N-terminus binding
          arachidonic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P102197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18999 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P102197 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P102197 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P102197 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P102197 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55825 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29921 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10760003 - -
          Affymetrix Rat Gene 1.0 ST Array 10760004 - -
          Affymetrix Rat Gene 1.0 ST Array 10760001 - -
          Affymetrix Rat Gene 1.0 ST Array 10760002 - -
          Affymetrix Rat Gene 1.0 ST Array 10760000 - -
          Affymetrix Rat Gene 1.1 ST Array 10759999 - -
          Rat Genome 230 2.0 Array 1369672_PM_at - -
          Rat Genome 230 2.0 Array 1369672_at - -
          Affymetrix HT Rat Focus Array 1369672_PM_at - -
          Affymetrix HT Rat Focus Array 1369672_at - -
          Affymetrix Rat Clariom S Assay PSR1200008638.rn.1 TC1200001002.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008636.rn.1 TC1200001002.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008634.rn.1 TC1200001002.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008637.rn.1 TC1200001002.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008635.rn.1 TC1200001002.rn.1 RTA TC
          Rat Expression Set 230 1369672_PM_at - -
          Rat Expression Set 230 1369672_at - -
          Affymetrix Rat Gene 1.0 ST Array 10759999 - -
          Rat Genome U34 Set rc_AI104781_at - -
          Rat Genome U34 Set X52196cds_at - -
          Rat Genome U34 Set rc_AA963933_i_at - -
          Rat Genome U34 Set rc_AA963933_f_at - -
          Rat Genome U34 Set X52196cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22352789 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357059 - -
          RatRef-12 Expression BeadChip ILMN_67910 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0