Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568718_m1
          See other PSMA6 GE Assays ›
          Gene Symbol:
          Psma6
          Gene Name:
          proteasome (prosome, macropain) subunit, alpha type 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 76174462 - 76205429 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568718_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psma6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017283.3 NP_058979.1 5 - 6 654 116
          XM_006240098.2 XP_006240160.1 5 - 6 612 116
          GenBank mRNA BC062232.1 - 5 - 6 655 6888689 116
          D10755.1 - 5 - 6 643 116
          FQ212808.1 - 756 116
          FQ220193.1 - 5 - 6 681 116

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psma6

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          positive regulation of NF-kappaB transcription factor activity
          proteolysis involved in cellular protein catabolic process
          RNA binding
          threonine-type endopeptidase activity
          NF-kappaB binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26964 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148947 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14214 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14214 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14214 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1006334 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14214 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19080 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10759496 - -
          Affymetrix Rat Gene 1.0 ST Array 10884659 - -
          Affymetrix Rat Gene 1.0 ST Array 10884662 - -
          Affymetrix Rat Gene 1.0 ST Array 10884658 - -
          Affymetrix Rat Gene 1.0 ST Array 10884660 - -
          Affymetrix Rat Gene 1.0 ST Array 10884657 - -
          Affymetrix Rat Gene 1.0 ST Array 10884661 - -
          Affymetrix Rat Gene 1.1 ST Array 10884656 - -
          Affymetrix Rat Gene 1.1 ST Array 10759495 - -
          Rat Genome 230 2.0 Array 1369930_PM_at - -
          Rat Genome 230 2.0 Array 1369930_at - -
          Affymetrix HT Rat Focus Array 1369930_PM_at - -
          Affymetrix HT Rat Focus Array 1369930_at - -
          Affymetrix Rat Clariom S Assay PSR0600005317.rn.1 TC0600000792.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600005316.rn.1 TC0600000792.rn.1 RTA TC
          Rat Expression Set 230 1369930_PM_at - -
          Rat Expression Set 230 1369930_at - -
          Affymetrix Rat Gene 1.0 ST Array 10759495 - -
          Affymetrix Rat Gene 1.0 ST Array 10884656 - -
          Rat Genome U34 Set rc_AA799492_at - -
          Rat Genome U34 Set D10755_s_at - -
          Rat Genome U34 Set rc_AI112633_s_at - -
          Rat Genome U34 Set rc_AI071208_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21204330 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370284 - -
          RatRef-12 Expression BeadChip ILMN_62273 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline