Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568721_m1
          See other PSMB2 GE Assays ›
          Gene Symbol:
          Psmb2
          Gene Name:
          proteasome (prosome, macropain) subunit, beta type 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 144702004 - 144734220 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568721_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017284.1 NP_058980.1 3 - 4 339 59
          GenBank mRNA BC058487.1 - 3 - 4 359 6891616 59
          D21799.1 - 3 - 4 339 59
          FQ217737.1 - 3 - 4 369 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001479.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmb2

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          response to organonitrogen compound
          response to organic cyclic compound
          proteolysis involved in cellular protein catabolic process
          molecular_function
          threonine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P577458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176052 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90474 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90357 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143750 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P577458 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P577458 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P577458 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P577458 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700178381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43252 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101838 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61240 - -
          Agilent Rat cDNA Microarray (G4105A) 700178381 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872068 - -
          Affymetrix Rat Gene 1.0 ST Array 10872064 - -
          Affymetrix Rat Gene 1.0 ST Array 10872065 - -
          Affymetrix Rat Gene 1.0 ST Array 10872063 - -
          Affymetrix Rat Gene 1.0 ST Array 10872066 - -
          Affymetrix Rat Gene 1.0 ST Array 10872067 - -
          Affymetrix Rat Gene 1.1 ST Array 10872062 - -
          Rat Genome 230 2.0 Array 1398786_at - -
          Rat Genome 230 2.0 Array 1398786_PM_at - -
          Affymetrix HT Rat Focus Array 1398786_PM_at - -
          Affymetrix HT Rat Focus Array 1398786_at - -
          Affymetrix Rat Clariom S Assay PSR0500010437.rn.1 TC0500001479.rn.1 RTA TC
          Rat Expression Set 230 1398786_at - -
          Rat Expression Set 230 1398786_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872062 - -
          Rat Genome U34 Set D21799_at - -
          Rat Genome U34 Set D21799_g_at - -
          Rat Genome U34 Set D21799_g_at - -
          Rat Genome U34 Set D21799_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21341028 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353270 - -
          RatRef-12 Expression BeadChip ILMN_58076 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline