Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568786_m1
          See other UHMK1 GE Assays ›
          Gene Symbol:
          Uhmk1
          Gene Name:
          U2AF homology motif (UHM) kinase 1
          Gene Aliases:
          KIS, Kist
          Chromosome Location:
          Chr.13: 88521625 - 88536728 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568786_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Uhmk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017293.1 NP_058989.1 3 - 4 909 135
          GenBank mRNA U70372.2 - 3 - 4 909 135
          X98374.1 - 3 - 4 786 135

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Uhmk1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle arrest
          peptidyl-serine phosphorylation
          neuron projection development
          positive regulation of translational initiation
          protein autophosphorylation
          regulation of protein export from nucleus
          nucleic acid binding
          RNA binding
          protein serine/threonine kinase activity
          ATP binding
          enzyme binding
          ribonucleoprotein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P165466 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P380743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110171 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P165466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P380743 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P380743 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P165466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P165466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P380743 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P513148 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11899 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702079186 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75515 - -
          Agilent Rat cDNA Microarray (G4105A) 702079186 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769722 - -
          Affymetrix Rat Gene 1.0 ST Array 10765461 - -
          Affymetrix Rat Gene 1.0 ST Array 10798470 - -
          Affymetrix Rat Gene 1.0 ST Array 10769720 - -
          Affymetrix Rat Gene 1.0 ST Array 10769721 - -
          Affymetrix Rat Gene 1.1 ST Array 10765460 - -
          Affymetrix Rat Gene 1.1 ST Array 10769719 - -
          Affymetrix Rat Gene 1.1 ST Array 10798469 - -
          Rat Genome 230 2.0 Array 1387406_PM_at - -
          Rat Genome 230 2.0 Array 1387406_at - -
          Affymetrix HT Rat Focus Array 1387406_at - -
          Affymetrix HT Rat Focus Array 1387406_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300013314.rn.1 TC1300001877.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013319.rn.1 TC1300001877.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013320.rn.1 TC1300001877.rn.1 RTA TC
          Rat Expression Set 230 1387406_PM_at - -
          Rat Expression Set 230 1387406_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769719 - -
          Affymetrix Rat Gene 1.0 ST Array 10798469 - -
          Rat Genome U34 Set U70372_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21363575 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358356 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355767 - -
          RatRef-12 Expression BeadChip ILMN_60180 - -
          RatRef-12 Expression BeadChip ILMN_58250 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline