Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568907_m1
          See other ECI1 GE Assays ›
          Gene Symbol:
          Eci1
          Gene Name:
          enoyl-CoA delta isomerase 1
          Gene Aliases:
          Dci
          Chromosome Location:
          Chr.10: 13797573 - 13810918 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568907_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eci1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017306.4 NP_059002.2 2 - 3 133 64
          GenBank mRNA BC078705.1 - 2 - 3 133 7129389 64
          FQ213384.1 - 2 - 3 141 64
          FQ218351.1 - 2 - 3 134 64
          M61112.1 - 2 - 3 122 64
          X61184.1 - 2 - 3 144 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000206.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eci1

          Panther Classification:

          Molecular Function -

          hydratase

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid beta-oxidation
          metabolic process
          dodecenoyl-CoA delta-isomerase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P337294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P337294 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P337294 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P337294 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P337294 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700481210 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25741 - -
          Agilent Rat cDNA Microarray (G4105A) 700481210 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10732075 - -
          Affymetrix Rat Gene 1.0 ST Array 10732071 - -
          Affymetrix Rat Gene 1.0 ST Array 10732072 - -
          Affymetrix Rat Gene 1.0 ST Array 10732073 - -
          Affymetrix Rat Gene 1.0 ST Array 10732074 - -
          Affymetrix Rat Gene 1.0 ST Array 10732070 - -
          Affymetrix Rat Gene 1.0 ST Array 10732069 - -
          Affymetrix Rat Gene 1.1 ST Array 10732068 - -
          Rat Genome 230 2.0 Array 1370961_at - -
          Rat Genome 230 2.0 Array 1370961_PM_at - -
          Rat Genome 230 2.0 Array 1392684_at - -
          Rat Genome 230 2.0 Array 1367659_s_at - -
          Rat Genome 230 2.0 Array 1392684_PM_at - -
          Rat Genome 230 2.0 Array 1367659_PM_s_at - -
          Affymetrix HT Rat Focus Array 1392684_PM_at - -
          Affymetrix HT Rat Focus Array 1370961_at - -
          Affymetrix HT Rat Focus Array 1370961_PM_at - -
          Affymetrix HT Rat Focus Array 1367659_PM_s_at - -
          Affymetrix HT Rat Focus Array 1367659_s_at - -
          Affymetrix HT Rat Focus Array 1392684_at - -
          Affymetrix Rat Clariom S Assay PSR1000001878.rn.1 TC1000000206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000001875.rn.1 TC1000000206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000001877.rn.1 TC1000000206.rn.1 RTA TC
          Rat Expression Set 230 1367659_s_at - -
          Rat Expression Set 230 1367659_PM_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10732068 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21592236 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361402 - -
          RatRef-12 Expression BeadChip ILMN_56804 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline