Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568971_m1
          See other SLC23A1 GE Assays ›
          Gene Symbol:
          Slc23a1
          Gene Name:
          solute carrier family 23 (ascorbic acid transporter), member 1
          Gene Aliases:
          SVCT1
          Chromosome Location:
          Chr.18: 28414008 - 28428151 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568971_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc23a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017315.2 NP_059011.1 14 - 15 1844 77
          XM_006254602.2 XP_006254664.1 16 - 17 2225 77
          GenBank mRNA AF080452.1 - 14 - 15 1844 77
          BC078851.1 - 14 - 15 1844 7127602 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001246.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc23a1

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          brain development
          response to toxic substance
          L-ascorbic acid transport
          lung development
          vitamin transmembrane transport
          sodium ion transmembrane transport
          dehydroascorbic acid transport
          transepithelial L-ascorbic acid transport
          transporter activity
          L-ascorbate:sodium symporter activity
          sodium ion transmembrane transporter activity
          L-ascorbic acid transporter activity
          dehydroascorbic acid transporter activity
          sodium-dependent L-ascorbate transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P823322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71839 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P823322 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P823322 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P823322 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P823322 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701563121 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102848 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73114 - -
          Agilent Rat cDNA Microarray (G4105A) 701563121 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803886 - -
          Affymetrix Rat Gene 1.0 ST Array 10803882 - -
          Affymetrix Rat Gene 1.0 ST Array 10803885 - -
          Affymetrix Rat Gene 1.0 ST Array 10803892 - -
          Affymetrix Rat Gene 1.0 ST Array 10803879 - -
          Affymetrix Rat Gene 1.0 ST Array 10803887 - -
          Affymetrix Rat Gene 1.0 ST Array 10803883 - -
          Affymetrix Rat Gene 1.0 ST Array 10803880 - -
          Affymetrix Rat Gene 1.0 ST Array 10803881 - -
          Affymetrix Rat Gene 1.0 ST Array 10803890 - -
          Affymetrix Rat Gene 1.0 ST Array 10803884 - -
          Affymetrix Rat Gene 1.0 ST Array 10803893 - -
          Affymetrix Rat Gene 1.0 ST Array 10803889 - -
          Affymetrix Rat Gene 1.0 ST Array 10803894 - -
          Affymetrix Rat Gene 1.0 ST Array 10803888 - -
          Affymetrix Rat Gene 1.0 ST Array 10803891 - -
          Affymetrix Rat Gene 1.1 ST Array 10803878 - -
          Rat Genome 230 2.0 Array 1369169_PM_at - -
          Rat Genome 230 2.0 Array 1369169_at - -
          Affymetrix HT Rat Focus Array 1369169_PM_at - -
          Affymetrix HT Rat Focus Array 1369169_at - -
          Affymetrix Rat Clariom S Assay PSR1800008855.rn.1 TC1800001246.rn.1 RTA TC
          Rat Expression Set 230 1369169_PM_at - -
          Rat Expression Set 230 1369169_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803878 - -
          Rat Genome U34 Set rc_AA996842_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21615614 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375024 - -
          RatRef-12 Expression BeadChip ILMN_63974 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline