Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569036_m1
          See other CTSS GE Assays ›
          Gene Symbol:
          Ctss
          Gene Name:
          cathepsin S
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 197655780 - 197679768 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569036_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctss
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017320.1 NP_059016.1 5 - 6 922 91
          GenBank mRNA FQ218804.1 - 997 91
          L03201.1 - 5 - 6 922 91

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctss

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          metanephros development
          adaptive immune response
          proteolysis
          collagen catabolic process
          basement membrane disassembly
          bone resorption
          positive regulation of inflammatory response
          proteolysis involved in cellular protein catabolic process
          regulation of sensory perception of pain
          cellular response to thyroid hormone stimulus
          fibronectin binding
          cysteine-type endopeptidase activity
          collagen binding
          peptidase activity
          cysteine-type peptidase activity
          laminin binding
          proteoglycan binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P355928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8739 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85650 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P355928 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P355928 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P355928 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1004731 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P355928 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700252861 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31273 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86553 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43446 - -
          Agilent Rat cDNA Microarray (G4105A) 700252861 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10817435 - -
          Affymetrix Rat Gene 1.0 ST Array 10817433 - -
          Affymetrix Rat Gene 1.0 ST Array 10817431 - -
          Affymetrix Rat Gene 1.0 ST Array 10817434 - -
          Affymetrix Rat Gene 1.0 ST Array 10817436 - -
          Affymetrix Rat Gene 1.0 ST Array 10817437 - -
          Affymetrix Rat Gene 1.0 ST Array 10817432 - -
          Affymetrix Rat Gene 1.1 ST Array 10817429 - -
          Rat Genome 230 2.0 Array 1387005_PM_at - -
          Rat Genome 230 2.0 Array 1387005_at - -
          Affymetrix HT Rat Focus Array 1387005_at - -
          Affymetrix HT Rat Focus Array 1387005_PM_at - -
          Rat Expression Set 230 1387005_at - -
          Rat Expression Set 230 1387005_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10817429 - -
          Rat Genome U34 Set L03201_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22001662 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352236 - -
          RatRef-12 Expression BeadChip ILMN_63508 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline