Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569175_m1
          See other PTPRO GE Assays ›
          Gene Symbol:
          Ptpro
          Gene Name:
          protein tyrosine phosphatase, receptor type, O
          Gene Aliases:
          GLEPP1, RPTP-BK
          Chromosome Location:
          Chr.4: 171251243 - 171461569 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569175_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptpro
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017336.1 NP_059032.1 23 - 24 3318 70
          XM_006237571.2 XP_006237633.1 12 - 13 1226 70
          XM_006237572.2 XP_006237634.1 11 - 12 1140 70
          GenBank mRNA D30749.1 - 1 - 2 121 70
          D45412.1 - 14 - 15 1557 70
          U28938.1 - 23 - 24 3318 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001968.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptpro

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          monocyte chemotaxis
          regulation of glomerular filtration
          negative regulation of glomerular filtration
          nervous system development
          axon guidance
          lamellipodium assembly
          glomerulus development
          peptidyl-tyrosine dephosphorylation
          slit diaphragm assembly
          glomerular visceral epithelial cell differentiation
          negative regulation of canonical Wnt signaling pathway
          protein tyrosine phosphatase activity
          Wnt-protein binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P289080 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P100008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704625 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80703 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15324 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147615 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77214 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125739 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152388 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167142 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130267 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112356 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58557 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87705 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134064 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49565 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70153 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P289080 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P289080 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P289080 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P100008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P289080 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P430937 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702024473 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45043 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29913 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13309 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40929 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96091 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17293 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61887 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104298 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1368 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30705 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90561 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5627 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40399 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91626 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58802 - -
          Agilent Rat cDNA Microarray (G4105A) 702024473 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10859352 - -
          Affymetrix Rat Gene 1.0 ST Array 10859358 - -
          Affymetrix Rat Gene 1.0 ST Array 10859359 - -
          Affymetrix Rat Gene 1.0 ST Array 10859353 - -
          Affymetrix Rat Gene 1.0 ST Array 10859345 - -
          Affymetrix Rat Gene 1.0 ST Array 10859364 - -
          Affymetrix Rat Gene 1.0 ST Array 10859356 - -
          Affymetrix Rat Gene 1.0 ST Array 10859367 - -
          Affymetrix Rat Gene 1.0 ST Array 10859348 - -
          Affymetrix Rat Gene 1.0 ST Array 10859365 - -
          Affymetrix Rat Gene 1.0 ST Array 10859361 - -
          Affymetrix Rat Gene 1.0 ST Array 10859366 - -
          Affymetrix Rat Gene 1.0 ST Array 10859349 - -
          Affymetrix Rat Gene 1.0 ST Array 10859360 - -
          Affymetrix Rat Gene 1.0 ST Array 10859343 - -
          Affymetrix Rat Gene 1.0 ST Array 10859355 - -
          Affymetrix Rat Gene 1.0 ST Array 10859350 - -
          Affymetrix Rat Gene 1.0 ST Array 10859347 - -
          Affymetrix Rat Gene 1.0 ST Array 10859368 - -
          Affymetrix Rat Gene 1.0 ST Array 10859370 - -
          Affymetrix Rat Gene 1.0 ST Array 10859354 - -
          Affymetrix Rat Gene 1.0 ST Array 10859346 - -
          Affymetrix Rat Gene 1.0 ST Array 10859351 - -
          Affymetrix Rat Gene 1.0 ST Array 10859357 - -
          Affymetrix Rat Gene 1.0 ST Array 10859363 - -
          Affymetrix Rat Gene 1.0 ST Array 10859344 - -
          Affymetrix Rat Gene 1.0 ST Array 10859362 - -
          Affymetrix Rat Gene 1.1 ST Array 10859342 - -
          Rat Genome 230 2.0 Array 1368412_PM_a_at - -
          Rat Genome 230 2.0 Array 1368412_a_at - -
          Affymetrix HT Rat Focus Array 1368412_a_at - -
          Affymetrix HT Rat Focus Array 1368412_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0400014158.rn.1 TC0400001968.rn.1 RTA TC
          Rat Expression Set 230 1368412_PM_a_at - -
          Rat Expression Set 230 1368412_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10859342 - -
          Rat Genome U34 Set D45412_s_at - -
          Rat Genome U34 Set U28938_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22128943 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349137 - -
          RatRef-12 Expression BeadChip ILMN_52068 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline