Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569224_m1
          See other LIPF GE Assays ›
          Gene Symbol:
          Lipf
          Gene Name:
          lipase, gastric
          Gene Aliases:
          RNLIP, Rnlp
          Chromosome Location:
          Chr.1: 252284464 - 252302811 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569224_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lipf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017341.2 NP_059037.1 9 - 10 1003 93
          GenBank mRNA A01157.1 - 8 - 9 962 93
          AA998080.1 - 1 - 2 93
          X02309.1 - 8 - 9 1004 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003505.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lipf

          Panther Classification:

          Molecular Function -

          hydrolase lipase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          malate metabolic process
          digestion
          lipid catabolic process
          oxidation-reduction process
          triglyceride lipase activity
          lipase activity
          malate dehydrogenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P004017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598605 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82300 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156279 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004017 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004017 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P004017 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11927 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7183 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102021 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10714855 - -
          Affymetrix Rat Gene 1.0 ST Array 10714852 - -
          Affymetrix Rat Gene 1.0 ST Array 10714849 - -
          Affymetrix Rat Gene 1.0 ST Array 10714850 - -
          Affymetrix Rat Gene 1.0 ST Array 10714853 - -
          Affymetrix Rat Gene 1.0 ST Array 10714856 - -
          Affymetrix Rat Gene 1.0 ST Array 10714854 - -
          Affymetrix Rat Gene 1.0 ST Array 10714857 - -
          Affymetrix Rat Gene 1.0 ST Array 10714851 - -
          Affymetrix Rat Gene 1.1 ST Array 10714848 - -
          Rat Genome 230 2.0 Array 1387245_PM_at - -
          Rat Genome 230 2.0 Array 1387245_at - -
          Affymetrix HT Rat Focus Array 1387245_PM_at - -
          Affymetrix HT Rat Focus Array 1387245_at - -
          Affymetrix Rat Clariom S Assay PSR0100028396.rn.1 TC0100003505.rn.1 RTA TC
          Rat Expression Set 230 1387245_PM_at - -
          Rat Expression Set 230 1387245_at - -
          Affymetrix Rat Gene 1.0 ST Array 10714848 - -
          Rat Genome U34 Set X02309_at - -
          Rat Genome U34 Set A01157cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21683531 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362989 - -
          RatRef-12 Expression BeadChip ILMN_52756 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline