Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569232_m1
          See other GSK3A GE Assays ›
          Gene Symbol:
          Gsk3a
          Gene Name:
          glycogen synthase kinase 3 alpha
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 82097244 - 82108203 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569232_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gsk3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017344.1 NP_059040.1 1 - 2 414 66
          XM_008758972.1 XP_008757194.1 1 - 2 402 66
          GenBank mRNA X53427.1 - 1 - 2 414 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004989.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus Rat Wnt Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gsk3a

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of systemic arterial blood pressure
          cardiac left ventricle morphogenesis
          glycogen metabolic process
          regulation of gene expression by genetic imprinting
          protein phosphorylation
          signal transduction
          nervous system development
          insulin receptor signaling pathway
          positive regulation of peptidyl-threonine phosphorylation
          Wnt signaling pathway
          cell migration
          positive regulation of cAMP biosynthetic process
          negative regulation of TOR signaling
          cellular response to insulin stimulus
          positive regulation of peptidyl-serine phosphorylation
          cellular response to interleukin-3
          proteasome-mediated ubiquitin-dependent protein catabolic process
          hypermethylation of CpG island
          positive regulation of heart contraction
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          negative regulation of transferase activity
          negative regulation of cell growth involved in cardiac muscle cell development
          cellular response to lithium ion
          cellular response to organic cyclic compound
          positive regulation of adrenergic receptor signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
          positive regulation of glycogen (starch) synthase activity
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          protein kinase A catalytic subunit binding
          tau-protein kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127609 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092255 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092255 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092255 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P276402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10621 - -
          Agilent Rat cDNA Microarray (G4105A) 700784720 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719803 - -
          Affymetrix Rat Gene 1.0 ST Array 10719802 - -
          Affymetrix Rat Gene 1.0 ST Array 10719801 - -
          Affymetrix Rat Gene 1.0 ST Array 10719807 - -
          Affymetrix Rat Gene 1.0 ST Array 10719804 - -
          Affymetrix Rat Gene 1.0 ST Array 10719799 - -
          Affymetrix Rat Gene 1.0 ST Array 10719806 - -
          Affymetrix Rat Gene 1.0 ST Array 10719800 - -
          Affymetrix Rat Gene 1.0 ST Array 10719805 - -
          Affymetrix Rat Gene 1.0 ST Array 10719808 - -
          Affymetrix Rat Gene 1.1 ST Array 10719798 - -
          Rat Genome 230 2.0 Array 1389714_PM_at - -
          Rat Genome 230 2.0 Array 1389714_at - -
          Rat Genome 230 2.0 Array 1386883_at - -
          Rat Genome 230 2.0 Array 1386883_PM_at - -
          Affymetrix HT Rat Focus Array 1386883_at - -
          Affymetrix HT Rat Focus Array 1386883_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100039600.rn.1 TC0100004989.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100039599.rn.1 TC0100004989.rn.1 RTA TC
          Rat Expression Set 230 1386883_PM_at - -
          Rat Expression Set 230 1386883_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719798 - -
          Rat Genome U34 Set X53427_at - -
          Rat Genome U34 Set X53427_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21155072 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376822 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline