Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569313_m1
          See other SLC7A5 GE Assays ›
          Gene Symbol:
          Slc7a5
          Gene Name:
          solute carrier family 7 (amino acid transporter light chain, L system), member 5
          Gene Aliases:
          E16, TA1
          Chromosome Location:
          Chr.19: 54693959 - 54722563 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569313_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc7a5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017353.1 NP_059049.1 1 - 2 605 77
          GenBank mRNA AB015432.1 - 1 - 2 605 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc7a5

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          neutral amino acid transport
          L-amino acid transport
          cell differentiation
          L-alpha-amino acid transmembrane transport
          amino acid transmembrane transporter activity
          L-amino acid transmembrane transporter activity
          peptide antigen binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P380575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532833 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120063 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P380575 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P380575 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P380575 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P380575 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701311729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41697 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62662 - -
          Agilent Rat cDNA Microarray (G4105A) 701311729 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811542 - -
          Affymetrix Rat Gene 1.0 ST Array 10811539 - -
          Affymetrix Rat Gene 1.0 ST Array 10811533 - -
          Affymetrix Rat Gene 1.0 ST Array 10811534 - -
          Affymetrix Rat Gene 1.0 ST Array 10811541 - -
          Affymetrix Rat Gene 1.0 ST Array 10811536 - -
          Affymetrix Rat Gene 1.0 ST Array 10811532 - -
          Affymetrix Rat Gene 1.0 ST Array 10811535 - -
          Affymetrix Rat Gene 1.0 ST Array 10811538 - -
          Affymetrix Rat Gene 1.0 ST Array 10811540 - -
          Affymetrix Rat Gene 1.0 ST Array 10811537 - -
          Affymetrix Rat Gene 1.1 ST Array 10811531 - -
          Rat Genome 230 2.0 Array 1387280_PM_a_at - -
          Rat Genome 230 2.0 Array 1387280_a_at - -
          Affymetrix HT Rat Focus Array 1387280_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387280_a_at - -
          Affymetrix Rat Clariom S Assay PSR1900013023.rn.1 TC1900001618.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013024.rn.1 TC1900001618.rn.1 RTA TC
          Rat Expression Set 230 1387280_PM_a_at - -
          Rat Expression Set 230 1387280_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811531 - -
          Rat Genome U34 Set AB015432_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21952815 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365729 - -
          RatRef-12 Expression BeadChip ILMN_56541 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline