Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569508_g1
          See other AVPR2 GE Assays ›
          Gene Symbol:
          Avpr2
          Gene Name:
          arginine vasopressin receptor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 156889006 - 156892646 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569508_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Avpr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019136.1 NP_062009.1 2 - 3 910 89
          XM_006229549.2 XP_006229611.1 3 - 4 2304 89
          GenBank mRNA AY338195.1 - 2 - 3 910 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Avpr2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of systemic arterial blood pressure by vasopressin
          positive regulation of systemic arterial blood pressure
          G-protein coupled receptor signaling pathway
          I-kappaB kinase/NF-kappaB signaling
          positive regulation of cell proliferation
          positive regulation of gene expression
          telencephalon development
          interferon-gamma production
          cellular response to hormone stimulus
          response to cytokine
          negative regulation of urine volume
          negative regulation of renal sodium excretion
          positive regulation of blood pressure
          positive regulation of vasoconstriction
          response to peptide
          vasopressin receptor activity
          peptide hormone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P031890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031890 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031890 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P031890 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11944 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96920 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936024 - -
          Affymetrix Rat Gene 1.0 ST Array 10936023 - -
          Affymetrix Rat Gene 1.0 ST Array 10936022 - -
          Affymetrix Rat Gene 1.1 ST Array 10936021 - -
          Affymetrix Rat Gene 1.1 ST Array 10940224 - -
          Rat Genome 230 2.0 Array 1369873_at - -
          Rat Genome 230 2.0 Array 1369873_PM_at - -
          Rat Genome 230 2.0 Array 1393958_at - -
          Rat Genome 230 2.0 Array 1393958_PM_at - -
          Affymetrix HT Rat Focus Array 1369873_PM_at - -
          Affymetrix HT Rat Focus Array 1369873_at - -
          Affymetrix Rat Clariom S Assay PSR0X00016445.rn.1 TC0X00002641.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016448.rn.1 TC0X00002641.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016446.rn.1 TC0X00002641.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00016447.rn.1 TC0X00002641.rn.1 RTA TC
          Rat Expression Set 230 1369873_PM_at - -
          Rat Expression Set 230 1369873_at - -
          Rat Expression Set 230 1393958_PM_at - -
          Rat Expression Set 230 1393958_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936021 - -
          Rat Genome U34 Set Z11932cds_at - -
          Rat Genome U34 Set Z11932cds_g_at - -
          Rat Genome U34 Set rc_AA819795_at - -
          Rat Genome U34 Set Z11932cds_g_at - -
          Rat Genome U34 Set Z11932cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21208685 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373218 - -
          RatRef-12 Expression BeadChip ILMN_66567 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline