Search
Search
| Gene Symbol: Acp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270889.1 | NP_001257818.1 | 3 - 4 | 543 | 95 | |
| NM_019144.2 | NP_062017.2 | 3 - 4 | 485 | 95 | ||
| XM_006242691.2 | XP_006242753.1 | 5 - 6 | 1191 | 95 | ||
| XM_006242692.2 | XP_006242754.1 | 4 - 5 | 1136 | 95 | ||
| XM_006242693.2 | XP_006242755.1 | 3 - 4 | 1002 | 95 | ||
| XM_006242694.2 | XP_006242756.1 | 3 - 4 | 439 | 95 | ||
| GenBank mRNA | BC078847.1 | - | 3 - 4 | 543 | 7110955 | 95 |
| CV074238.1 | - | 3 - 4 | 485 | 95 | ||
| M76110.1 | - | 3 - 4 | 483 | 95 | ||
Set Membership: |
|