Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn00569682_m1
See other UCN GE Assays ›
Gene Symbol:
Ucn
Gene Name:
urocortin
Gene Aliases:
-
Chromosome Location:
Chr.6: 26602144 - 26602974 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn00569682_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
2,694.00
Your Price
Online offer:
2694.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Ucn
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_019150.1
NP_062023.1
1 - 2
48
61
GenBank mRNA
U33935.1
-
1 - 2
48
61
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_019150.1, U33935.1 transcripts.
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
3' Most
Detect Genomic DNA
Rattus norvegicus
1700
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Ucn
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
inflammatory response
response to oxidative stress
neuropeptide signaling pathway
female pregnancy
sensory perception of sound
learning or memory
feeding behavior
associative learning
aerobic respiration
positive regulation of gene expression
negative regulation of gene expression
pancreatic juice secretion
positive regulation of cell growth
positive regulation of cAMP biosynthetic process
negative regulation of histone deacetylation
neuron projection development
negative regulation of appetite
response to estradiol
positive regulation of interleukin-6 production
positive regulation of collagen biosynthetic process
positive regulation of peptidyl-serine phosphorylation
activation of protein kinase A activity
social behavior
gastric emptying
drinking behavior
negative regulation of apoptotic process
positive regulation of vascular permeability
positive regulation of translation
positive regulation of DNA replication
negative regulation of blood pressure
negative regulation of cell size
positive regulation of vasodilation
negative regulation of hormone secretion
response to pain
response to glucocorticoid
positive regulation of corticotropin secretion
regulation of synaptic transmission, glutamatergic
positive regulation of cardiac muscle contraction
negative regulation of gastric acid secretion
negative regulation of necrotic cell death
negative regulation of cell death
positive regulation of calcium ion import
negative regulation of neuron death
negative regulation of feeding behavior
positive regulation of behavioral fear response
G-protein coupled receptor binding
hormone activity
protein binding
histone deacetylase inhibitor activity
corticotropin-releasing hormone receptor 1 binding
corticotropin-releasing hormone receptor 2 binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_64_P058336
-
-
Agilent SurePrint G3 Rat CGH Microarray 1x1M
154261
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P058336
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P058336
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_64_P058336
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_43_P11951
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Gene 1.0 ST Array
10883160
-
-
Affymetrix Rat Gene 1.0 ST Array
10883161
-
-
Affymetrix Rat Gene 1.1 ST Array
10883159
-
-
Rat Genome 230 2.0 Array
1387645_at
-
-
Rat Genome 230 2.0 Array
1387645_PM_at
-
-
Affymetrix HT Rat Focus Array
1387645_at
-
-
Affymetrix HT Rat Focus Array
1387645_PM_at
-
-
Rat Expression Set 230
1387645_at
-
-
Rat Expression Set 230
1387645_PM_at
-
-
Affymetrix Rat Gene 1.0 ST Array
10883159
-
-
Rat Genome U34 Set
U33935_at
-
-
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
21413383
-
-
Array Name
Probe IDs
Targets
Target Type
Illumina ratRef-12 v1.0 expression beadchip
ILMN_1360395
-
-
RatRef-12 Expression BeadChip
ILMN_49013
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline