Search
Search
| Gene Symbol: Aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019157.2 | NP_062030.2 | 2 - 3 | 362 | 82 | |
| GenBank mRNA | AB000507.2 | - | 2 - 3 | 362 | 82 | |
| AY120935.1 | - | 1 - 2 | 101 | 82 | ||
| AY157737.1 | - | 1 - 2 | 101 | 82 | ||
| BC072695.1 | - | 2 - 3 | 322 | 7104684 | 82 | |
Set Membership: |
|