Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569745_m1
          See other UTS2 GE Assays ›
          Gene Symbol:
          Uts2
          Gene Name:
          urotensin 2
          Gene Aliases:
          Ucn2
          Chromosome Location:
          Chr.5: 168078748 - 168084133 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569745_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Uts2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019160.2 NP_062033.1 1 - 2 141 73
          GenBank mRNA AF172174.1 - 1 - 2 141 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001972.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Uts2

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          negative regulation of glomerular filtration
          positive regulation of cytosolic calcium ion concentration
          negative regulation of heart rate
          positive regulation of heart rate
          positive regulation of fibroblast migration
          positive regulation of circadian sleep/wake cycle, wakefulness
          positive regulation of synaptic transmission, cholinergic
          positive regulation of collagen biosynthetic process
          response to testosterone
          negative regulation of urine volume
          negative regulation of renal sodium excretion
          response to drug
          positive regulation of cell differentiation
          positive regulation of angiogenesis
          negative regulation of blood pressure
          positive regulation of blood pressure
          positive regulation of vasodilation
          positive regulation of circadian sleep/wake cycle, REM sleep
          negative regulation of insulin secretion
          positive regulation of fibroblast proliferation
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P078020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078020 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P078020 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P531908 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102444 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10874213 - -
          Affymetrix Rat Gene 1.0 ST Array 10874215 - -
          Affymetrix Rat Gene 1.0 ST Array 10874214 - -
          Affymetrix Rat Gene 1.0 ST Array 10874212 - -
          Affymetrix Rat Gene 1.1 ST Array 10874210 - -
          Rat Genome 230 2.0 Array 1368805_at - -
          Rat Genome 230 2.0 Array 1368805_PM_at - -
          Affymetrix HT Rat Focus Array 1368805_at - -
          Affymetrix HT Rat Focus Array 1368805_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500014810.rn.1 TC0500001972.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500014809.rn.1 TC0500001972.rn.1 RTA TC
          Rat Expression Set 230 1368805_at - -
          Rat Expression Set 230 1368805_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10874210 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21862177 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372205 - -
          RatRef-12 Expression BeadChip ILMN_61193 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline