Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569763_m1
          See other PSEN1 GE Assays ›
          Gene Symbol:
          Psen1
          Gene Name:
          presenilin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 107169513 - 107220957 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569763_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psen1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019163.3 NP_062036.2 2 - 3 338 101
          XM_006240321.2 XP_006240383.1 4 - 5 608 101
          XM_006240322.2 XP_006240384.1 4 - 5 861 101
          GenBank mRNA BC070887.1 - 4 - 5 534 7098018 101
          D82363.1 - 2 - 3 338 101
          D82578.1 - 2 - 3 338 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001144.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psen1

          Panther Classification:

          Molecular Function -

          aspartic protease

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPKK activity
          blood vessel development
          cell fate specification
          somitogenesis
          neuron migration
          positive regulation of receptor recycling
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          heart looping
          hematopoietic progenitor cell differentiation
          T cell activation involved in immune response
          myeloid leukocyte differentiation
          neural retina development
          negative regulation of protein kinase activity
          protein glycosylation
          proteolysis
          membrane protein ectodomain proteolysis
          calcium ion transport
          mitochondrial transport
          cellular calcium ion homeostasis
          autophagy
          cellular response to DNA damage stimulus
          response to oxidative stress
          negative regulation of epidermal growth factor-activated receptor activity
          regulation of epidermal growth factor-activated receptor activity
          Notch signaling pathway
          Notch receptor processing
          brain development
          heart development
          learning or memory
          memory
          post-embryonic development
          protein transport
          L-glutamate transport
          choline transport
          synaptic vesicle targeting
          single organismal cell-cell adhesion
          protein processing
          cerebral cortex cell migration
          Cajal-Retzius cell differentiation
          dorsal/ventral neural tube patterning
          cerebral cortex development
          neurogenesis
          neuron differentiation
          embryonic limb morphogenesis
          forebrain development
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          endoplasmic reticulum calcium ion homeostasis
          beta-amyloid formation
          segmentation
          intracellular signal transduction
          locomotion
          regulation of phosphorylation
          eye photoreceptor cell development
          amyloid precursor protein catabolic process
          myeloid dendritic cell differentiation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          regulation of protein binding
          positive regulation of MAP kinase activity
          negative regulation of neuron apoptotic process
          skin morphogenesis
          cellular protein metabolic process
          positive regulation of protein kinase activity
          regulation of synaptic plasticity
          thymus development
          neuron development
          skeletal system morphogenesis
          brain morphogenesis
          beta-amyloid metabolic process
          epithelial cell proliferation
          negative regulation of axonogenesis
          positive regulation of coagulation
          T cell receptor signaling pathway
          neuron apoptotic process
          negative regulation of ubiquitin-protein transferase activity
          smooth endoplasmic reticulum calcium ion homeostasis
          protein maturation
          regulation of synaptic transmission, glutamatergic
          canonical Wnt signaling pathway
          regulation of resting membrane potential
          calcium ion transmembrane transport
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic signaling pathway
          endopeptidase activity
          aspartic-type endopeptidase activity
          calcium channel activity
          protein binding
          beta-catenin binding
          peptidase activity
          PDZ domain binding
          cadherin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P044315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93150 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104559 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59092 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044315 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044315 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P044315 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P142016 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700485743 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11548 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1819 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50151 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94525 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31649 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13930 - -
          Agilent Rat cDNA Microarray (G4105A) 700485743 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10885813 - -
          Affymetrix Rat Gene 1.0 ST Array 10885818 - -
          Affymetrix Rat Gene 1.0 ST Array 10885814 - -
          Affymetrix Rat Gene 1.0 ST Array 10885810 - -
          Affymetrix Rat Gene 1.0 ST Array 10885819 - -
          Affymetrix Rat Gene 1.0 ST Array 10885812 - -
          Affymetrix Rat Gene 1.0 ST Array 10885817 - -
          Affymetrix Rat Gene 1.0 ST Array 10885820 - -
          Affymetrix Rat Gene 1.0 ST Array 10885821 - -
          Affymetrix Rat Gene 1.0 ST Array 10885809 - -
          Affymetrix Rat Gene 1.0 ST Array 10885811 - -
          Affymetrix Rat Gene 1.0 ST Array 10885816 - -
          Affymetrix Rat Gene 1.0 ST Array 10885822 - -
          Affymetrix Rat Gene 1.0 ST Array 10885815 - -
          Affymetrix Rat Gene 1.1 ST Array 10885808 - -
          Rat Genome 230 2.0 Array 1376075_at - -
          Rat Genome 230 2.0 Array 1368792_PM_at - -
          Rat Genome 230 2.0 Array 1368792_at - -
          Rat Genome 230 2.0 Array 1376075_PM_at - -
          Affymetrix HT Rat Focus Array 1376075_PM_at - -
          Affymetrix HT Rat Focus Array 1376075_at - -
          Affymetrix HT Rat Focus Array 1368792_PM_at - -
          Affymetrix HT Rat Focus Array 1368792_at - -
          Affymetrix Rat Clariom S Assay PSR0600007959.rn.1 TC0600001144.rn.1 RTA TC
          Rat Expression Set 230 1368792_at - -
          Rat Expression Set 230 1376075_at - -
          Rat Expression Set 230 1368792_PM_at - -
          Rat Expression Set 230 1376075_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10885808 - -
          Rat Genome U34 Set D82363_s_at - -
          Rat Genome U34 Set rc_AI009495_at - -
          Rat Genome U34 Set D82363_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21150134 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351184 - -
          RatRef-12 Expression BeadChip ILMN_57702 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline